Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6651751-6651984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf100
Entrez Gene ID
388327
Gene Name
chromosome 17 open reading frame 100
Gene Aliases
-
UniGene
Hs.262480
Gene Chromosome Location
Chr.17: 6651739-6653298 on Build GRCh38
Transcripts
NM_001105520.1 XM_017024710.1

Gene Symbol
MED31
Entrez Gene ID
51003
Gene Name
mediator complex subunit 31
Gene Aliases
3110004H13Rik, CGI-125, Soh1
UniGene
Hs.567493
Gene Chromosome Location
Chr.17: 6643313-6651804 on Build GRCh38
Transcripts
NM_001105520.1 XM_017024710.1

SNP Detail

SNP ID Chromosome Location
rs550444095 Chr.17: 6651833-6651833 on Build GRCh38
rs749242374 Chr.17: 6651964-6651964 on Build GRCh38
rs571052314 Chr.17: 6651946-6651946 on Build GRCh38
rs528105937 Chr.17: 6651817-6651817 on Build GRCh38
rs1033660584 Chr.17: 6651799-6651799 on Build GRCh38
rs560875922 Chr.17: 6651875-6651875 on Build GRCh38
rs372262464 Chr.17: 6651777-6651777 on Build GRCh38
rs149318341 Chr.17: 6651898-6651898 on Build GRCh38
rs73351900 Chr.17: 6651905-6651905 on Build GRCh38
rs766894052 Chr.17: 6651892-6651892 on Build GRCh38
rs758806477 Chr.17: 6651932-6651932 on Build GRCh38
rs1014014713 Chr.17: 6651948-6651948 on Build GRCh38
rs756314754 Chr.17: 6651870-6651870 on Build GRCh38
rs779442321 Chr.17: 6651907-6651907 on Build GRCh38
rs747190510 Chr.17: 6651945-6651945 on Build GRCh38
rs945489497 Chr.17: 6651864-6651864 on Build GRCh38
rs981038413 Chr.17: 6651781-6651781 on Build GRCh38
rs1023259208 Chr.17: 6651863-6651863 on Build GRCh38
rs961390909 Chr.17: 6651931-6651931 on Build GRCh38
rs922459959 Chr.17: 6651887-6651887 on Build GRCh38
rs528939179 Chr.17: 6651822-6651822 on Build GRCh38
rs1032226647 Chr.17: 6651831-6651831 on Build GRCh38
rs941061467 Chr.17: 6651927-6651927 on Build GRCh38
rs990788575 Chr.17: 6651848-6651848 on Build GRCh38
rs763511439 Chr.17: 6651895-6651895 on Build GRCh38
rs566183978 Chr.17: 6651937-6651937 on Build GRCh38
rs928304833 Chr.17: 6651908-6651908 on Build GRCh38
rs750777843 Chr.17: 6651926-6651926 on Build GRCh38
rs375365664 Chr.17: 6651778-6651778 on Build GRCh38
rs545336606 Chr.17: 6651801-6651801 on Build GRCh38
rs781074770 Chr.17: 6651950-6651950 on Build GRCh38
rs760292575 Chr.17: 6651893-6651893 on Build GRCh38
rs573264152 Chr.17: 6651794-6651794 on Build GRCh38

View Full Product Details