Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173359062-173359335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA7
Entrez Gene ID
83879
Gene Name
cell division cycle associated 7
Gene Aliases
ICF3, JPO1
UniGene
Hs.470654
Gene Chromosome Location
Chr.2: 173354017-173368992 on Build GRCh38
Transcripts
NM_031942.4 NM_145810.2 XM_011511957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1613711 c.148-4A>G p.? Chr.2: 173359251-173359251 on Build GRCh38
7158660 c.191T>C p.V64A Chr.2: 173359298-173359298 on Build GRCh38
2893807 c.225C>T p.C75C Chr.2: 173359332-173359332 on Build GRCh38
5394965 c.218C>T p.S73F Chr.2: 173359325-173359325 on Build GRCh38
6624520 c.226G>A p.G76S Chr.2: 173359333-173359333 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766590861 Chr.2: 173359302-173359302 on Build GRCh38
rs769639621 Chr.2: 173359234-173359237 on Build GRCh38
rs948855860 Chr.2: 173359220-173359220 on Build GRCh38
rs982705167 Chr.2: 173359157-173359157 on Build GRCh38
rs772970252 Chr.2: 173359297-173359298 on Build GRCh38
rs768414403 Chr.2: 173359215-173359215 on Build GRCh38
rs984220260 Chr.2: 173359284-173359284 on Build GRCh38
rs867990868 Chr.2: 173359303-173359303 on Build GRCh38
rs768919236 Chr.2: 173359146-173359146 on Build GRCh38
rs572399327 Chr.2: 173359187-173359187 on Build GRCh38
rs767086066 Chr.2: 173359269-173359269 on Build GRCh38
rs199813398 Chr.2: 173359232-173359232 on Build GRCh38
rs752605887 Chr.2: 173359275-173359275 on Build GRCh38
rs542096054 Chr.2: 173359185-173359185 on Build GRCh38
rs763915331 Chr.2: 173359289-173359289 on Build GRCh38
rs747882390 Chr.2: 173359215-173359216 on Build GRCh38
rs781675874 Chr.2: 173359212-173359212 on Build GRCh38
rs773208652 Chr.2: 173359194-173359194 on Build GRCh38
rs774145403 Chr.2: 173359243-173359243 on Build GRCh38
rs934860592 Chr.2: 173359311-173359311 on Build GRCh38
rs955284756 Chr.2: 173359155-173359155 on Build GRCh38
rs748642254 Chr.2: 173359221-173359221 on Build GRCh38
rs760707630 Chr.2: 173359279-173359279 on Build GRCh38
rs369933460 Chr.2: 173359258-173359258 on Build GRCh38
rs753722248 Chr.2: 173359291-173359291 on Build GRCh38
rs144464133 Chr.2: 173359171-173359171 on Build GRCh38
rs974156628 Chr.2: 173359192-173359192 on Build GRCh38
rs556932741 Chr.2: 173359137-173359137 on Build GRCh38
rs776499818 Chr.2: 173359215-173359217 on Build GRCh38
rs867081577 Chr.2: 173359276-173359276 on Build GRCh38
rs879773060 Chr.2: 173359102-173359102 on Build GRCh38
rs770090468 Chr.2: 173359223-173359223 on Build GRCh38
rs757038223 Chr.2: 173359296-173359296 on Build GRCh38
rs952424682 Chr.2: 173359231-173359231 on Build GRCh38
rs773670717 Chr.2: 173359229-173359229 on Build GRCh38
rs889390595 Chr.2: 173359163-173359163 on Build GRCh38
rs75662905 Chr.2: 173359186-173359186 on Build GRCh38
rs747003176 Chr.2: 173359213-173359214 on Build GRCh38

View Full Product Details