Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157690300-157690565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
UniGene
Hs.292449
Gene Chromosome Location
Chr.1: 157676481-157700985 on Build GRCh38
Transcripts
NM_001320333.1 NM_052939.3 NR_135214.1 NR_135215.1 NR_135216.1 NR_135217.1 XM_006711145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334193 c.1443G>T p.R481S Chr.1: 157690502-157690502 on Build GRCh38
3863068 c.1515G>A p.P505P Chr.1: 157690430-157690430 on Build GRCh38
6583616 c.1480G>C p.E494Q Chr.1: 157690465-157690465 on Build GRCh38
5700097 c.1514C>A p.P505Q Chr.1: 157690431-157690431 on Build GRCh38
6058508 c.1606A>T p.T536S Chr.1: 157690339-157690339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765815493 Chr.1: 157690474-157690474 on Build GRCh38
rs139158249 Chr.1: 157690529-157690529 on Build GRCh38
rs760761491 Chr.1: 157690404-157690404 on Build GRCh38
rs763368387 Chr.1: 157690475-157690475 on Build GRCh38
rs143597458 Chr.1: 157690495-157690495 on Build GRCh38
rs375740832 Chr.1: 157690347-157690347 on Build GRCh38
rs922057447 Chr.1: 157690524-157690524 on Build GRCh38
rs754435989 Chr.1: 157690492-157690492 on Build GRCh38
rs1047864428 Chr.1: 157690500-157690500 on Build GRCh38
rs756043575 Chr.1: 157690443-157690443 on Build GRCh38
rs990630432 Chr.1: 157690441-157690441 on Build GRCh38
rs780154078 Chr.1: 157690516-157690516 on Build GRCh38
rs749114025 Chr.1: 157690501-157690501 on Build GRCh38
rs773784322 Chr.1: 157690530-157690530 on Build GRCh38
rs768838705 Chr.1: 157690462-157690462 on Build GRCh38
rs757010354 Chr.1: 157690511-157690511 on Build GRCh38
rs772310432 Chr.1: 157690398-157690398 on Build GRCh38
rs776846309 Chr.1: 157690469-157690469 on Build GRCh38
rs772208010 Chr.1: 157690458-157690458 on Build GRCh38
rs750783956 Chr.1: 157690428-157690428 on Build GRCh38
rs762396625 Chr.1: 157690414-157690414 on Build GRCh38
rs543072365 Chr.1: 157690468-157690468 on Build GRCh38
rs1020637077 Chr.1: 157690361-157690361 on Build GRCh38
rs943107070 Chr.1: 157690465-157690465 on Build GRCh38
rs143813895 Chr.1: 157690528-157690528 on Build GRCh38
rs371736717 Chr.1: 157690438-157690438 on Build GRCh38
rs375088342 Chr.1: 157690493-157690493 on Build GRCh38
rs148588983 Chr.1: 157690437-157690437 on Build GRCh38
rs752173870 Chr.1: 157690494-157690494 on Build GRCh38
rs144058263 Chr.1: 157690461-157690461 on Build GRCh38
rs767160539 Chr.1: 157690521-157690521 on Build GRCh38
rs529239679 Chr.1: 157690348-157690348 on Build GRCh38
rs147268020 Chr.1: 157690431-157690431 on Build GRCh38
rs369176005 Chr.1: 157690408-157690408 on Build GRCh38
rs143115777 Chr.1: 157690365-157690365 on Build GRCh38
rs775741944 Chr.1: 157690459-157690459 on Build GRCh38
rs772982173 Chr.1: 157690382-157690382 on Build GRCh38
rs201980342 Chr.1: 157690420-157690420 on Build GRCh38
rs777588617 Chr.1: 157690448-157690448 on Build GRCh38
rs369593542 Chr.1: 157690522-157690522 on Build GRCh38
rs200237068 Chr.1: 157690359-157690359 on Build GRCh38
rs765948607 Chr.1: 157690478-157690478 on Build GRCh38
rs765762115 Chr.1: 157690422-157690422 on Build GRCh38
rs747937607 Chr.1: 157690374-157690374 on Build GRCh38
rs762447964 Chr.1: 157690471-157690471 on Build GRCh38
rs138291587 Chr.1: 157690430-157690430 on Build GRCh38
rs768497919 Chr.1: 157690363-157690363 on Build GRCh38
rs775482909 Chr.1: 157690402-157690402 on Build GRCh38
rs777639956 Chr.1: 157690496-157690496 on Build GRCh38
rs1024818888 Chr.1: 157690455-157690455 on Build GRCh38
rs759823787 Chr.1: 157690383-157690383 on Build GRCh38
rs6691220 Chr.1: 157690480-157690480 on Build GRCh38
rs151156070 Chr.1: 157690456-157690456 on Build GRCh38
rs141302640 Chr.1: 157690409-157690409 on Build GRCh38
rs149831176 Chr.1: 157690432-157690432 on Build GRCh38
rs769525001 Chr.1: 157690379-157690379 on Build GRCh38

View Full Product Details