Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196994195-196994426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR5
Entrez Gene ID
81494
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
UniGene
Hs.282594
Gene Chromosome Location
Chr.1: 196975022-197009725 on Build GRCh38
Transcripts
NM_030787.3 XM_011510020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5029596 c.679+5G>C p.? Chr.1: 196994261-196994261 on Build GRCh38
1560359 c.588T>G p.P196P Chr.1: 196994237-196994237 on Build GRCh38
3480943 c.623C>T p.S208L Chr.1: 196994200-196994200 on Build GRCh38
3480945 c.651G>A p.G217G Chr.1: 196994228-196994228 on Build GRCh38
6123871 c.607G>T p.G203* Chr.1: 196994256-196994256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758984179 Chr.1: 196994231-196994231 on Build GRCh38
rs1020994350 Chr.1: 196994280-196994280 on Build GRCh38
rs772108485 Chr.1: 196994295-196994295 on Build GRCh38
rs772906571 Chr.1: 196994272-196994272 on Build GRCh38
rs765907289 Chr.1: 196994307-196994307 on Build GRCh38
rs996323734 Chr.1: 196994393-196994393 on Build GRCh38
rs564711982 Chr.1: 196994309-196994309 on Build GRCh38
rs138105988 Chr.1: 196994243-196994243 on Build GRCh38
rs746120951 Chr.1: 196994277-196994277 on Build GRCh38
rs760845759 Chr.1: 196994334-196994334 on Build GRCh38
rs318240755 Chr.1: 196994232-196994232 on Build GRCh38
rs757424508 Chr.1: 196994248-196994248 on Build GRCh38
rs781352172 Chr.1: 196994256-196994256 on Build GRCh38
rs919959310 Chr.1: 196994275-196994275 on Build GRCh38
rs912344774 Chr.1: 196994333-196994333 on Build GRCh38
rs534590754 Chr.1: 196994328-196994328 on Build GRCh38
rs762820494 Chr.1: 196994291-196994291 on Build GRCh38
rs964741951 Chr.1: 196994392-196994392 on Build GRCh38
rs765183774 Chr.1: 196994233-196994234 on Build GRCh38
rs773149761 Chr.1: 196994297-196994297 on Build GRCh38
rs765656585 Chr.1: 196994223-196994223 on Build GRCh38
rs769856245 Chr.1: 196994281-196994281 on Build GRCh38
rs146361151 Chr.1: 196994294-196994294 on Build GRCh38
rs754586890 Chr.1: 196994222-196994222 on Build GRCh38
rs552166236 Chr.1: 196994282-196994282 on Build GRCh38
rs760541432 Chr.1: 196994300-196994300 on Build GRCh38
rs981688848 Chr.1: 196994234-196994234 on Build GRCh38
rs778137947 Chr.1: 196994240-196994240 on Build GRCh38
rs553438814 Chr.1: 196994367-196994367 on Build GRCh38
rs151134004 Chr.1: 196994225-196994225 on Build GRCh38
rs1013913757 Chr.1: 196994250-196994250 on Build GRCh38
rs1039585812 Chr.1: 196994381-196994381 on Build GRCh38
rs958682898 Chr.1: 196994257-196994257 on Build GRCh38
rs190137007 Chr.1: 196994260-196994260 on Build GRCh38

View Full Product Details