Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5753730-5753985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
UniGene
Hs.646899
Gene Chromosome Location
Chr.4: 5711197-5829043 on Build GRCh38
Transcripts
NM_001306090.1 NM_001306092.1 NM_153717.2 XM_006713865.3 XM_006713866.3 XM_011513419.2 XM_017007883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6656815 c.1436C>T p.P479L Chr.4: 5753905-5753905 on Build GRCh38
447976 c.1412G>A p.R471K Chr.4: 5753881-5753881 on Build GRCh38
3302184 c.1449G>A p.P483P Chr.4: 5753918-5753918 on Build GRCh38
4987948 c.1328G>A p.R443Q Chr.4: 5753797-5753797 on Build GRCh38
5785514 c.1436C>G p.P479R Chr.4: 5753905-5753905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375242825 Chr.4: 5753892-5753892 on Build GRCh38
rs752575646 Chr.4: 5753819-5753819 on Build GRCh38
rs886038740 Chr.4: 5753953-5753953 on Build GRCh38
rs933791675 Chr.4: 5753948-5753948 on Build GRCh38
rs753296851 Chr.4: 5753945-5753945 on Build GRCh38
rs369734950 Chr.4: 5753938-5753938 on Build GRCh38
rs765596595 Chr.4: 5753858-5753858 on Build GRCh38
rs779553464 Chr.4: 5753878-5753878 on Build GRCh38
rs372236822 Chr.4: 5753885-5753885 on Build GRCh38
rs767660467 Chr.4: 5753760-5753760 on Build GRCh38
rs986329571 Chr.4: 5753859-5753859 on Build GRCh38
rs35953626 Chr.4: 5753797-5753797 on Build GRCh38
rs968007063 Chr.4: 5753816-5753816 on Build GRCh38
rs146232611 Chr.4: 5753931-5753931 on Build GRCh38
rs764643954 Chr.4: 5753873-5753873 on Build GRCh38
rs775819388 Chr.4: 5753918-5753918 on Build GRCh38
rs770992412 Chr.4: 5753909-5753909 on Build GRCh38
rs1036703453 Chr.4: 5753776-5753776 on Build GRCh38
rs376440801 Chr.4: 5753917-5753917 on Build GRCh38
rs773408319 Chr.4: 5753836-5753836 on Build GRCh38
rs116236414 Chr.4: 5753941-5753941 on Build GRCh38
rs748062945 Chr.4: 5753829-5753829 on Build GRCh38
rs573106140 Chr.4: 5753755-5753755 on Build GRCh38
rs202150959 Chr.4: 5753790-5753790 on Build GRCh38
rs763393820 Chr.4: 5753868-5753868 on Build GRCh38
rs768485560 Chr.4: 5753885-5753885 on Build GRCh38
rs772689168 Chr.4: 5753757-5753757 on Build GRCh38
rs753064604 Chr.4: 5753865-5753865 on Build GRCh38
rs112861104 Chr.4: 5753906-5753906 on Build GRCh38
rs753152908 Chr.4: 5753793-5753793 on Build GRCh38
rs141859946 Chr.4: 5753838-5753838 on Build GRCh38
rs201393812 Chr.4: 5753792-5753792 on Build GRCh38
rs116952023 Chr.4: 5753802-5753802 on Build GRCh38
rs754316550 Chr.4: 5753924-5753924 on Build GRCh38
rs759736593 Chr.4: 5753915-5753915 on Build GRCh38
rs773461223 Chr.4: 5753765-5753765 on Build GRCh38
rs896913430 Chr.4: 5753778-5753778 on Build GRCh38
rs745395865 Chr.4: 5753814-5753814 on Build GRCh38
rs60582583 Chr.4: 5753789-5753789 on Build GRCh38
rs765877009 Chr.4: 5753943-5753943 on Build GRCh38
rs753483489 Chr.4: 5753888-5753888 on Build GRCh38
rs777731204 Chr.4: 5753961-5753961 on Build GRCh38
rs778599838 Chr.4: 5753827-5753827 on Build GRCh38
rs778770904 Chr.4: 5753957-5753957 on Build GRCh38
rs571637567 Chr.4: 5753897-5753897 on Build GRCh38
rs774390669 Chr.4: 5753754-5753754 on Build GRCh38
rs760918290 Chr.4: 5753769-5753769 on Build GRCh38
rs772145574 Chr.4: 5753831-5753831 on Build GRCh38
rs924563658 Chr.4: 5753907-5753907 on Build GRCh38
rs754641764 Chr.4: 5753956-5753956 on Build GRCh38
rs755691949 Chr.4: 5753876-5753876 on Build GRCh38
rs758314449 Chr.4: 5753960-5753960 on Build GRCh38
rs752505533 Chr.4: 5753958-5753958 on Build GRCh38
rs748009991 Chr.4: 5753905-5753905 on Build GRCh38
rs377753181 Chr.4: 5753963-5753963 on Build GRCh38
rs766679717 Chr.4: 5753785-5753785 on Build GRCh38
rs372893834 Chr.4: 5753908-5753908 on Build GRCh38
rs778787222 Chr.4: 5753901-5753901 on Build GRCh38
rs202205463 Chr.4: 5753852-5753852 on Build GRCh38
rs138725840 Chr.4: 5753846-5753846 on Build GRCh38
rs749008952 Chr.4: 5753818-5753818 on Build GRCh38
rs977322984 Chr.4: 5753903-5753903 on Build GRCh38
rs764556070 Chr.4: 5753932-5753932 on Build GRCh38
rs760607210 Chr.4: 5753840-5753840 on Build GRCh38
rs944576189 Chr.4: 5753898-5753898 on Build GRCh38
rs2302075 Chr.4: 5753815-5753815 on Build GRCh38
rs146028983 Chr.4: 5753845-5753845 on Build GRCh38

View Full Product Details