Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42306467-42306704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANC
Entrez Gene ID
2595
Gene Name
glucosidase alpha, neutral C
Gene Aliases
-
UniGene
Hs.730806
Gene Chromosome Location
Chr.15: 42273658-42353666 on Build GRCh38
Transcripts
NM_001301409.1 NM_198141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4796794 c.577G>A p.G193S Chr.15: 42306564-42306564 on Build GRCh38
4686389 c.604delT p.F202fs*79 Chr.15: 42306591-42306591 on Build GRCh38
961694 c.604T>C p.F202L Chr.15: 42306591-42306591 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141667276 Chr.15: 42306507-42306507 on Build GRCh38
rs898004434 Chr.15: 42306548-42306548 on Build GRCh38
rs992551783 Chr.15: 42306614-42306614 on Build GRCh38
rs776355247 Chr.15: 42306608-42306608 on Build GRCh38
rs370343407 Chr.15: 42306642-42306642 on Build GRCh38
rs772322658 Chr.15: 42306596-42306596 on Build GRCh38
rs151186877 Chr.15: 42306589-42306589 on Build GRCh38
rs749957880 Chr.15: 42306505-42306505 on Build GRCh38
rs781468452 Chr.15: 42306530-42306530 on Build GRCh38
rs754637901 Chr.15: 42306527-42306527 on Build GRCh38
rs761719912 Chr.15: 42306553-42306555 on Build GRCh38
rs201067273 Chr.15: 42306500-42306500 on Build GRCh38
rs748510035 Chr.15: 42306539-42306539 on Build GRCh38
rs370302153 Chr.15: 42306650-42306650 on Build GRCh38
rs1018128696 Chr.15: 42306630-42306630 on Build GRCh38
rs746123321 Chr.15: 42306594-42306594 on Build GRCh38
rs778198915 Chr.15: 42306546-42306546 on Build GRCh38
rs766088478 Chr.15: 42306518-42306518 on Build GRCh38
rs917004899 Chr.15: 42306667-42306667 on Build GRCh38
rs773234152 Chr.15: 42306618-42306618 on Build GRCh38
rs762954548 Chr.15: 42306619-42306619 on Build GRCh38
rs756620033 Chr.15: 42306542-42306542 on Build GRCh38
rs756425778 Chr.15: 42306663-42306663 on Build GRCh38
rs373238537 Chr.15: 42306632-42306632 on Build GRCh38
rs200142331 Chr.15: 42306525-42306525 on Build GRCh38
rs765226916 Chr.15: 42306582-42306582 on Build GRCh38
rs373118869 Chr.15: 42306580-42306580 on Build GRCh38
rs767133893 Chr.15: 42306651-42306651 on Build GRCh38
rs776541765 Chr.15: 42306530-42306530 on Build GRCh38
rs751069413 Chr.15: 42306645-42306645 on Build GRCh38
rs751210962 Chr.15: 42306526-42306526 on Build GRCh38
rs770954512 Chr.15: 42306586-42306586 on Build GRCh38
rs750389426 Chr.15: 42306613-42306614 on Build GRCh38
rs35753497 Chr.15: 42306676-42306676 on Build GRCh38
rs753076991 Chr.15: 42306658-42306658 on Build GRCh38
rs761898091 Chr.15: 42306499-42306499 on Build GRCh38
rs761632227 Chr.15: 42306616-42306616 on Build GRCh38

View Full Product Details