Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11824381-11824654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN6
Entrez Gene ID
1185
Gene Name
chloride voltage-gated channel 6
Gene Aliases
CLC-6
UniGene
Hs.193043
Gene Chromosome Location
Chr.1: 11806096-11843144 on Build GRCh38
Transcripts
NM_001256959.1 NM_001286.3 NR_046428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3472089 c.643C>T p.P215S Chr.1: 11824548-11824548 on Build GRCh38
5477525 c.581-2A>G p.? Chr.1: 11824484-11824484 on Build GRCh38
3472088 c.608T>C p.M203T Chr.1: 11824513-11824513 on Build GRCh38
6248173 c.609G>T p.M203I Chr.1: 11824514-11824514 on Build GRCh38
2179358 c.623C>T p.S208L Chr.1: 11824528-11824528 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779197982 Chr.1: 11824557-11824557 on Build GRCh38
rs761403516 Chr.1: 11824449-11824449 on Build GRCh38
rs547643862 Chr.1: 11824436-11824436 on Build GRCh38
rs995796351 Chr.1: 11824603-11824603 on Build GRCh38
rs748424449 Chr.1: 11824464-11824464 on Build GRCh38
rs774688041 Chr.1: 11824477-11824477 on Build GRCh38
rs771893561 Chr.1: 11824482-11824482 on Build GRCh38
rs767912902 Chr.1: 11824509-11824509 on Build GRCh38
rs781451847 Chr.1: 11824446-11824446 on Build GRCh38
rs74053322 Chr.1: 11824571-11824571 on Build GRCh38
rs368673791 Chr.1: 11824480-11824480 on Build GRCh38
rs746383670 Chr.1: 11824447-11824447 on Build GRCh38
rs377229046 Chr.1: 11824566-11824567 on Build GRCh38
rs772935956 Chr.1: 11824487-11824487 on Build GRCh38
rs374523990 Chr.1: 11824442-11824442 on Build GRCh38
rs746740245 Chr.1: 11824573-11824573 on Build GRCh38
rs41275474 Chr.1: 11824562-11824562 on Build GRCh38
rs749318511 Chr.1: 11824463-11824463 on Build GRCh38
rs750367442 Chr.1: 11824579-11824587 on Build GRCh38
rs764776408 Chr.1: 11824500-11824500 on Build GRCh38
rs139212698 Chr.1: 11824545-11824545 on Build GRCh38
rs138998252 Chr.1: 11824508-11824508 on Build GRCh38
rs770733916 Chr.1: 11824575-11824575 on Build GRCh38
rs768892949 Chr.1: 11824472-11824472 on Build GRCh38
rs772447262 Chr.1: 11824467-11824467 on Build GRCh38
rs754422676 Chr.1: 11824533-11824533 on Build GRCh38
rs776580850 Chr.1: 11824496-11824496 on Build GRCh38
rs201379697 Chr.1: 11824441-11824441 on Build GRCh38
rs375654164 Chr.1: 11824440-11824440 on Build GRCh38
rs149647864 Chr.1: 11824507-11824507 on Build GRCh38
rs371138196 Chr.1: 11824631-11824631 on Build GRCh38
rs975570351 Chr.1: 11824493-11824493 on Build GRCh38
rs973083278 Chr.1: 11824512-11824512 on Build GRCh38
rs113220356 Chr.1: 11824620-11824620 on Build GRCh38
rs750697542 Chr.1: 11824528-11824528 on Build GRCh38
rs112392420 Chr.1: 11824569-11824570 on Build GRCh38
rs759510987 Chr.1: 11824604-11824604 on Build GRCh38
rs200586664 Chr.1: 11824494-11824494 on Build GRCh38
rs772126084 Chr.1: 11824570-11824570 on Build GRCh38
rs1054049546 Chr.1: 11824437-11824437 on Build GRCh38
rs776527925 Chr.1: 11824599-11824599 on Build GRCh38
rs198400 Chr.1: 11824498-11824498 on Build GRCh38
rs569400909 Chr.1: 11824517-11824517 on Build GRCh38
rs1027181018 Chr.1: 11824618-11824618 on Build GRCh38
rs369655043 Chr.1: 11824434-11824434 on Build GRCh38
rs1033263713 Chr.1: 11824478-11824478 on Build GRCh38
rs369290855 Chr.1: 11824456-11824456 on Build GRCh38
rs1000193112 Chr.1: 11824627-11824627 on Build GRCh38
rs60602304 Chr.1: 11824529-11824529 on Build GRCh38

View Full Product Details