Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203305564-203305831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG2
Entrez Gene ID
7832
Gene Name
BTG anti-proliferation factor 2
Gene Aliases
APRO1, PC3, TIS21
UniGene
Hs.519162
Gene Chromosome Location
Chr.1: 203305536-203309602 on Build GRCh38
Transcripts
NM_006763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5714925 c.124C>G p.L42V Chr.1: 203305730-203305730 on Build GRCh38
5777122 c.11G>T p.G4V Chr.1: 203305617-203305617 on Build GRCh38
3774429 c.41C>T p.A14V Chr.1: 203305647-203305647 on Build GRCh38
28357 c.81G>A p.R27R Chr.1: 203305687-203305687 on Build GRCh38
7233633 c.142+6C>T p.? Chr.1: 203305754-203305754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372466316 Chr.1: 203305751-203305751 on Build GRCh38
rs891937219 Chr.1: 203305664-203305664 on Build GRCh38
rs781174805 Chr.1: 203305665-203305665 on Build GRCh38
rs756931202 Chr.1: 203305756-203305756 on Build GRCh38
rs567131728 Chr.1: 203305768-203305768 on Build GRCh38
rs754308829 Chr.1: 203305611-203305611 on Build GRCh38
rs892306063 Chr.1: 203305702-203305702 on Build GRCh38
rs769569694 Chr.1: 203305667-203305667 on Build GRCh38
rs909545329 Chr.1: 203305595-203305595 on Build GRCh38
rs764960689 Chr.1: 203305764-203305764 on Build GRCh38
rs553396231 Chr.1: 203305767-203305767 on Build GRCh38
rs745592014 Chr.1: 203305779-203305779 on Build GRCh38
rs951211630 Chr.1: 203305791-203305791 on Build GRCh38
rs772251147 Chr.1: 203305700-203305700 on Build GRCh38
rs765600993 Chr.1: 203305741-203305741 on Build GRCh38
rs777565959 Chr.1: 203305638-203305638 on Build GRCh38
rs530501274 Chr.1: 203305703-203305703 on Build GRCh38
rs745690867 Chr.1: 203305666-203305666 on Build GRCh38
rs754149177 Chr.1: 203305715-203305715 on Build GRCh38
rs759954930 Chr.1: 203305723-203305723 on Build GRCh38
rs975093188 Chr.1: 203305621-203305621 on Build GRCh38
rs906117398 Chr.1: 203305617-203305617 on Build GRCh38
rs909629661 Chr.1: 203305811-203305811 on Build GRCh38
rs920897695 Chr.1: 203305624-203305624 on Build GRCh38
rs190037595 Chr.1: 203305800-203305800 on Build GRCh38
rs752095028 Chr.1: 203305622-203305622 on Build GRCh38
rs746720055 Chr.1: 203305654-203305654 on Build GRCh38
rs6682806 Chr.1: 203305808-203305808 on Build GRCh38
rs55906353 Chr.1: 203305689-203305689 on Build GRCh38
rs766615672 Chr.1: 203305704-203305704 on Build GRCh38
rs749294784 Chr.1: 203305799-203305799 on Build GRCh38
rs770829293 Chr.1: 203305656-203305656 on Build GRCh38
rs56045415 Chr.1: 203305784-203305784 on Build GRCh38
rs1057478532 Chr.1: 203305660-203305660 on Build GRCh38
rs763748747 Chr.1: 203305753-203305753 on Build GRCh38
rs147064341 Chr.1: 203305616-203305616 on Build GRCh38
rs531950637 Chr.1: 203305612-203305612 on Build GRCh38
rs1010748312 Chr.1: 203305694-203305694 on Build GRCh38
rs551190656 Chr.1: 203305747-203305747 on Build GRCh38
rs567838091 Chr.1: 203305637-203305637 on Build GRCh38
rs753317706 Chr.1: 203305748-203305748 on Build GRCh38
rs756003273 Chr.1: 203305780-203305780 on Build GRCh38
rs755396120 Chr.1: 203305613-203305613 on Build GRCh38
rs766776936 Chr.1: 203305609-203305609 on Build GRCh38
rs761793365 Chr.1: 203305698-203305698 on Build GRCh38
rs368773937 Chr.1: 203305600-203305600 on Build GRCh38
rs775502657 Chr.1: 203305673-203305673 on Build GRCh38
rs1018238394 Chr.1: 203305746-203305746 on Build GRCh38
rs867116264 Chr.1: 203305649-203305649 on Build GRCh38
rs540175244 Chr.1: 203305760-203305760 on Build GRCh38
rs377192922 Chr.1: 203305761-203305761 on Build GRCh38

View Full Product Details