Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 185745444-185745685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
-
UniGene
Hs.98025
Gene Chromosome Location
Chr.2: 185738895-185833289 on Build GRCh38
Transcripts
NM_173651.3

SNP Detail

SNP ID Chromosome Location
rs775883059 Chr.2: 185745493-185745493 on Build GRCh38
rs1003693871 Chr.2: 185745602-185745602 on Build GRCh38
rs189396495 Chr.2: 185745537-185745537 on Build GRCh38
rs962166469 Chr.2: 185745632-185745632 on Build GRCh38
rs17813736 Chr.2: 185745661-185745661 on Build GRCh38
rs762047358 Chr.2: 185745555-185745555 on Build GRCh38
rs906304167 Chr.2: 185745553-185745553 on Build GRCh38
rs539029488 Chr.2: 185745520-185745520 on Build GRCh38
rs903337868 Chr.2: 185745475-185745475 on Build GRCh38
rs986055190 Chr.2: 185745663-185745663 on Build GRCh38
rs1053853606 Chr.2: 185745484-185745484 on Build GRCh38
rs184963416 Chr.2: 185745503-185745503 on Build GRCh38
rs1054691950 Chr.2: 185745526-185745526 on Build GRCh38
rs916287120 Chr.2: 185745496-185745496 on Build GRCh38
rs750339043 Chr.2: 185745616-185745616 on Build GRCh38
rs11884004 Chr.2: 185745617-185745617 on Build GRCh38
rs761435619 Chr.2: 185745536-185745536 on Build GRCh38
rs575968972 Chr.2: 185745541-185745541 on Build GRCh38
rs755540136 Chr.2: 185745552-185745552 on Build GRCh38
rs1027499194 Chr.2: 185745646-185745646 on Build GRCh38
rs893407324 Chr.2: 185745534-185745534 on Build GRCh38
rs765392511 Chr.2: 185745603-185745603 on Build GRCh38
rs993308356 Chr.2: 185745633-185745633 on Build GRCh38
rs779229570 Chr.2: 185745601-185745601 on Build GRCh38

View Full Product Details