Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182963978-182964217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D1
Entrez Gene ID
54165
Gene Name
defective in cullin neddylation 1 domain containing 1
Gene Aliases
DCNL1, DCUN1L1, RP42, SCCRO, SCRO, Tes3
UniGene
Hs.592789
Gene Chromosome Location
Chr.3: 182938158-182986364 on Build GRCh38
Transcripts
NM_001308101.1 NM_020640.3 XM_011512912.1 XM_011512914.1 XM_011512915.2 XM_011512916.2 XM_017006633.1 XM_017006634.1 XM_017006635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971931 c.221-1G>T p.? Chr.3: 182964050-182964050 on Build GRCh38
6373508 c.234G>T p.E78D Chr.3: 182964036-182964036 on Build GRCh38
7076981 c.226C>G p.Q76E Chr.3: 182964044-182964044 on Build GRCh38
4800470 c.223C>G p.P75A Chr.3: 182964047-182964047 on Build GRCh38
6260048 c.285C>A p.L95L Chr.3: 182963985-182963985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012172164 Chr.3: 182964172-182964172 on Build GRCh38
rs180816359 Chr.3: 182964161-182964161 on Build GRCh38
rs773454225 Chr.3: 182964039-182964039 on Build GRCh38
rs4859147 Chr.3: 182964065-182964065 on Build GRCh38
rs374689513 Chr.3: 182964047-182964047 on Build GRCh38
rs763048538 Chr.3: 182964046-182964046 on Build GRCh38
rs528558028 Chr.3: 182964149-182964149 on Build GRCh38
rs761288976 Chr.3: 182964064-182964064 on Build GRCh38
rs570860616 Chr.3: 182964143-182964143 on Build GRCh38
rs750109958 Chr.3: 182964081-182964081 on Build GRCh38
rs552281409 Chr.3: 182964103-182964103 on Build GRCh38
rs770191360 Chr.3: 182964022-182964022 on Build GRCh38
rs368282297 Chr.3: 182964095-182964095 on Build GRCh38
rs760671999 Chr.3: 182964060-182964060 on Build GRCh38
rs765918758 Chr.3: 182964090-182964090 on Build GRCh38
rs200467854 Chr.3: 182964128-182964128 on Build GRCh38
rs1003131580 Chr.3: 182964177-182964177 on Build GRCh38
rs772799259 Chr.3: 182964063-182964065 on Build GRCh38
rs375240677 Chr.3: 182964091-182964091 on Build GRCh38
rs200199547 Chr.3: 182964066-182964066 on Build GRCh38
rs764898784 Chr.3: 182964080-182964080 on Build GRCh38
rs1033368069 Chr.3: 182964182-182964182 on Build GRCh38
rs548809578 Chr.3: 182964059-182964059 on Build GRCh38
rs865834731 Chr.3: 182964170-182964170 on Build GRCh38

View Full Product Details