Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1010552-1010777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381161 c.1622T>A p.M541K Chr.19: 1010591-1010591 on Build GRCh38
6374663 c.1577C>A p.A526E Chr.19: 1010636-1010636 on Build GRCh38
6381722 c.1550C>T p.A517V Chr.19: 1010663-1010663 on Build GRCh38
6992894 c.1481A>T p.D494V Chr.19: 1010732-1010732 on Build GRCh38
5891532 c.1517C>T p.S506L Chr.19: 1010696-1010696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775848544 Chr.19: 1010599-1010599 on Build GRCh38
rs922430115 Chr.19: 1010727-1010727 on Build GRCh38
rs561681748 Chr.19: 1010659-1010659 on Build GRCh38
rs750517552 Chr.19: 1010639-1010639 on Build GRCh38
rs758058478 Chr.19: 1010660-1010660 on Build GRCh38
rs762153382 Chr.19: 1010635-1010635 on Build GRCh38
rs756897645 Chr.19: 1010725-1010725 on Build GRCh38
rs760855478 Chr.19: 1010695-1010695 on Build GRCh38
rs763859039 Chr.19: 1010710-1010710 on Build GRCh38
rs767581676 Chr.19: 1010636-1010636 on Build GRCh38
rs760663235 Chr.19: 1010644-1010644 on Build GRCh38
rs778709761 Chr.19: 1010657-1010657 on Build GRCh38
rs779554733 Chr.19: 1010747-1010747 on Build GRCh38
rs961578826 Chr.19: 1010713-1010713 on Build GRCh38
rs370807950 Chr.19: 1010755-1010755 on Build GRCh38
rs749847410 Chr.19: 1010682-1010682 on Build GRCh38
rs751530483 Chr.19: 1010610-1010610 on Build GRCh38
rs527744500 Chr.19: 1010691-1010691 on Build GRCh38
rs2240159 Chr.19: 1010692-1010692 on Build GRCh38
rs972546106 Chr.19: 1010637-1010637 on Build GRCh38
rs889461539 Chr.19: 1010658-1010658 on Build GRCh38
rs541213100 Chr.19: 1010700-1010700 on Build GRCh38
rs1050284409 Chr.19: 1010704-1010705 on Build GRCh38
rs183061118 Chr.19: 1010702-1010702 on Build GRCh38
rs746587856 Chr.19: 1010669-1010669 on Build GRCh38
rs1027470607 Chr.19: 1010575-1010575 on Build GRCh38
rs745319187 Chr.19: 1010737-1010737 on Build GRCh38
rs765100951 Chr.19: 1010701-1010701 on Build GRCh38
rs753995364 Chr.19: 1010581-1010581 on Build GRCh38
rs763461027 Chr.19: 1010601-1010601 on Build GRCh38
rs769114027 Chr.19: 1010611-1010611 on Build GRCh38
rs753693052 Chr.19: 1010653-1010653 on Build GRCh38
rs11540362 Chr.19: 1010674-1010674 on Build GRCh38
rs753874054 Chr.19: 1010696-1010696 on Build GRCh38
rs567001833 Chr.19: 1010746-1010746 on Build GRCh38
rs543289243 Chr.19: 1010651-1010651 on Build GRCh38
rs568738514 Chr.19: 1010729-1010729 on Build GRCh38
rs754805969 Chr.19: 1010656-1010656 on Build GRCh38
rs1044001211 Chr.19: 1010673-1010673 on Build GRCh38
rs994885629 Chr.19: 1010692-1010708 on Build GRCh38
rs910938492 Chr.19: 1010752-1010752 on Build GRCh38
rs201676598 Chr.19: 1010683-1010683 on Build GRCh38
rs780652243 Chr.19: 1010681-1010681 on Build GRCh38
rs952103278 Chr.19: 1010593-1010593 on Build GRCh38
rs556881874 Chr.19: 1010718-1010718 on Build GRCh38
rs531446822 Chr.19: 1010641-1010641 on Build GRCh38
rs566862093 Chr.19: 1010687-1010687 on Build GRCh38
rs1016879025 Chr.19: 1010712-1010712 on Build GRCh38
rs755550740 Chr.19: 1010740-1010740 on Build GRCh38
rs747604535 Chr.19: 1010639-1010662 on Build GRCh38
rs774820642 Chr.19: 1010629-1010629 on Build GRCh38

View Full Product Details