Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63550485-63550748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF7
Entrez Gene ID
10238
Gene Name
DDB1 and CUL4 associated factor 7
Gene Aliases
AN11, HAN11, SWAN-1, WDR68
UniGene
Hs.410596
Gene Chromosome Location
Chr.17: 63550435-63594279 on Build GRCh38
Transcripts
NM_005828.4 NR_073585.1

SNP Detail

SNP ID Chromosome Location
rs1014731927 Chr.17: 63550584-63550584 on Build GRCh38
rs943818336 Chr.17: 63550558-63550558 on Build GRCh38
rs772876029 Chr.17: 63550723-63550723 on Build GRCh38
rs746045356 Chr.17: 63550686-63550686 on Build GRCh38
rs867457265 Chr.17: 63550597-63550597 on Build GRCh38
rs756523119 Chr.17: 63550651-63550651 on Build GRCh38
rs1027693541 Chr.17: 63550625-63550625 on Build GRCh38
rs893424615 Chr.17: 63550617-63550617 on Build GRCh38
rs139146412 Chr.17: 63550546-63550546 on Build GRCh38
rs923681737 Chr.17: 63550575-63550575 on Build GRCh38
rs961228271 Chr.17: 63550516-63550516 on Build GRCh38
rs761821277 Chr.17: 63550710-63550710 on Build GRCh38
rs112793660 Chr.17: 63550656-63550656 on Build GRCh38
rs952729299 Chr.17: 63550541-63550541 on Build GRCh38
rs771164880 Chr.17: 63550663-63550663 on Build GRCh38
rs530493288 Chr.17: 63550604-63550604 on Build GRCh38
rs768561024 Chr.17: 63550694-63550694 on Build GRCh38
rs528274897 Chr.17: 63550512-63550512 on Build GRCh38
rs564651732 Chr.17: 63550555-63550555 on Build GRCh38
rs777836135 Chr.17: 63550652-63550652 on Build GRCh38
rs1003788878 Chr.17: 63550562-63550562 on Build GRCh38
rs762592029 Chr.17: 63550653-63550654 on Build GRCh38
rs780877170 Chr.17: 63550577-63550577 on Build GRCh38
rs567134371 Chr.17: 63550621-63550621 on Build GRCh38
rs935037701 Chr.17: 63550517-63550517 on Build GRCh38
rs1053519670 Chr.17: 63550520-63550520 on Build GRCh38
rs953414805 Chr.17: 63550646-63550646 on Build GRCh38
rs546982068 Chr.17: 63550698-63550698 on Build GRCh38
rs966299247 Chr.17: 63550664-63550664 on Build GRCh38
rs893497828 Chr.17: 63550522-63550522 on Build GRCh38
rs947698690 Chr.17: 63550528-63550528 on Build GRCh38
rs1012209519 Chr.17: 63550626-63550626 on Build GRCh38
rs559544793 Chr.17: 63550509-63550509 on Build GRCh38
rs765196521 Chr.17: 63550722-63550722 on Build GRCh38
rs748750368 Chr.17: 63550628-63550628 on Build GRCh38
rs375509503 Chr.17: 63550561-63550561 on Build GRCh38
rs919880593 Chr.17: 63550538-63550538 on Build GRCh38
rs550188568 Chr.17: 63550618-63550618 on Build GRCh38
rs759789614 Chr.17: 63550510-63550510 on Build GRCh38
rs774604572 Chr.17: 63550677-63550677 on Build GRCh38

View Full Product Details