Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35502727-35502998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMKN
Entrez Gene ID
93099
Gene Name
dermokine
Gene Aliases
UNQ729, ZD52F10
UniGene
Hs.417795
Gene Chromosome Location
Chr.19: 35497217-35513678 on Build GRCh38
Transcripts
NM_001126056.2 NM_001126059.2 NM_001190347.1 NM_001308380.1 NM_001308383.1 NM_033317.4 NR_033746.1 XM_006723477.1 XM_006723484.1 XM_006723489.1 XM_006723493.2 XM_006723494.2 XM_006723503.2 XM_011527494.2 XM_011527495.2 XM_011527496.2 XM_011527497.2 XM_011527498.2 XM_011527499.2 XM_011527500.2 XM_011527501.2 XM_011527502.2 XM_011527503.2 XM_011527504.2 XM_011527505.2 XM_011527506.2 XM_011527507.2 XM_011527508.2 XM_011527509.2 XM_011527510.2 XM_011527513.1 XM_011527514.1 XM_017027475.1 XM_017027476.1 XM_017027477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3796965 c.282C>T p.V94V Chr.19: 35502878-35502878 on Build GRCh38
6645625 c.1144C>T p.P382S Chr.19: 35502877-35502877 on Build GRCh38
5631155 c.1135C>T p.L379F Chr.19: 35502853-35502853 on Build GRCh38
3892313 c.289C>T p.P97S Chr.19: 35502871-35502871 on Build GRCh38
5464490 c.1135-1G>T p.? Chr.19: 35502887-35502887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780416458 Chr.19: 35502795-35502796 on Build GRCh38
rs537272766 Chr.19: 35502943-35502943 on Build GRCh38
rs764775732 Chr.19: 35502875-35502875 on Build GRCh38
rs369745403 Chr.19: 35502796-35502796 on Build GRCh38
rs777748419 Chr.19: 35502883-35502883 on Build GRCh38
rs1043832292 Chr.19: 35502856-35502856 on Build GRCh38
rs146956236 Chr.19: 35502863-35502863 on Build GRCh38
rs761196911 Chr.19: 35502920-35502920 on Build GRCh38
rs368351185 Chr.19: 35502812-35502812 on Build GRCh38
rs1048651539 Chr.19: 35502824-35502824 on Build GRCh38
rs374587394 Chr.19: 35502797-35502797 on Build GRCh38
rs201058202 Chr.19: 35502919-35502919 on Build GRCh38
rs1007625383 Chr.19: 35502868-35502868 on Build GRCh38
rs748058652 Chr.19: 35502831-35502831 on Build GRCh38
rs1026200463 Chr.19: 35502913-35502913 on Build GRCh38
rs766708531 Chr.19: 35502864-35502864 on Build GRCh38
rs774435982 Chr.19: 35502858-35502858 on Build GRCh38
rs375958975 Chr.19: 35502917-35502917 on Build GRCh38
rs888675909 Chr.19: 35502826-35502826 on Build GRCh38
rs984564462 Chr.19: 35502794-35502794 on Build GRCh38
rs199854660 Chr.19: 35502862-35502862 on Build GRCh38
rs753436058 Chr.19: 35502865-35502865 on Build GRCh38
rs115235751 Chr.19: 35502777-35502777 on Build GRCh38
rs371628447 Chr.19: 35502779-35502779 on Build GRCh38
rs556863512 Chr.19: 35502949-35502949 on Build GRCh38
rs778418778 Chr.19: 35502825-35502825 on Build GRCh38
rs958050291 Chr.19: 35502762-35502762 on Build GRCh38
rs750863663 Chr.19: 35502798-35502798 on Build GRCh38
rs768979175 Chr.19: 35502868-35502869 on Build GRCh38
rs749846262 Chr.19: 35502823-35502825 on Build GRCh38
rs775591210 Chr.19: 35502847-35502847 on Build GRCh38
rs772264102 Chr.19: 35502918-35502918 on Build GRCh38
rs374003971 Chr.19: 35502922-35502922 on Build GRCh38
rs1015909623 Chr.19: 35502936-35502937 on Build GRCh38
rs979813478 Chr.19: 35502944-35502944 on Build GRCh38
rs1039415093 Chr.19: 35502857-35502857 on Build GRCh38
rs377001986 Chr.19: 35502901-35502901 on Build GRCh38
rs766199260 Chr.19: 35502803-35502803 on Build GRCh38
rs768967630 Chr.19: 35502907-35502907 on Build GRCh38
rs768645449 Chr.19: 35502853-35502853 on Build GRCh38
rs756512290 Chr.19: 35502799-35502799 on Build GRCh38
rs777104075 Chr.19: 35502933-35502933 on Build GRCh38
rs761959444 Chr.19: 35502861-35502861 on Build GRCh38
rs754611162 Chr.19: 35502870-35502870 on Build GRCh38
rs757900991 Chr.19: 35502881-35502881 on Build GRCh38
rs749490476 Chr.19: 35502905-35502905 on Build GRCh38
rs771230741 Chr.19: 35502929-35502929 on Build GRCh38
rs147573481 Chr.19: 35502876-35502876 on Build GRCh38
rs758379666 Chr.19: 35502836-35502836 on Build GRCh38
rs777658209 Chr.19: 35502840-35502840 on Build GRCh38
rs371583115 Chr.19: 35502916-35502916 on Build GRCh38
rs746893178 Chr.19: 35502885-35502885 on Build GRCh38
rs566034279 Chr.19: 35502813-35502813 on Build GRCh38
rs11084816 Chr.19: 35502759-35502759 on Build GRCh38
rs774402317 Chr.19: 35502912-35502912 on Build GRCh38
rs368062269 Chr.19: 35502902-35502902 on Build GRCh38
rs749463512 Chr.19: 35502851-35502851 on Build GRCh38
rs746874546 Chr.19: 35502841-35502841 on Build GRCh38

View Full Product Details