Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8189649-8189890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS6
Entrez Gene ID
54785
Gene Name
BLOC-1 related complex subunit 6
Gene Aliases
C17orf59, PRO2472
UniGene
Hs.129563
Gene Chromosome Location
Chr.17: 8188333-8190246 on Build GRCh38
Transcripts
NM_017622.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708538 c.40G>A p.E14K Chr.17: 8189699-8189699 on Build GRCh38
5981140 c.458G>A p.S153N Chr.17: 8189683-8189683 on Build GRCh38
381147 c.389G>A p.G130E Chr.17: 8189752-8189752 on Build GRCh38
1147918 c.442G>A p.E148K Chr.17: 8189699-8189699 on Build GRCh38
4070976 c.421G>C p.E141Q Chr.17: 8189720-8189720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769105399 Chr.17: 8189739-8189739 on Build GRCh38
rs773853354 Chr.17: 8189811-8189811 on Build GRCh38
rs370859187 Chr.17: 8189742-8189742 on Build GRCh38
rs751749586 Chr.17: 8189831-8189831 on Build GRCh38
rs758504849 Chr.17: 8189775-8189775 on Build GRCh38
rs759498783 Chr.17: 8189755-8189755 on Build GRCh38
rs1033079371 Chr.17: 8189697-8189697 on Build GRCh38
rs144247363 Chr.17: 8189720-8189720 on Build GRCh38
rs533276759 Chr.17: 8189854-8189854 on Build GRCh38
rs750447631 Chr.17: 8189774-8189774 on Build GRCh38
rs781400698 Chr.17: 8189797-8189797 on Build GRCh38
rs757505255 Chr.17: 8189693-8189693 on Build GRCh38
rs772963022 Chr.17: 8189803-8189803 on Build GRCh38
rs754812108 Chr.17: 8189671-8189671 on Build GRCh38
rs374315817 Chr.17: 8189745-8189745 on Build GRCh38
rs748434754 Chr.17: 8189851-8189851 on Build GRCh38
rs748615756 Chr.17: 8189800-8189800 on Build GRCh38
rs905857681 Chr.17: 8189682-8189682 on Build GRCh38
rs778027585 Chr.17: 8189856-8189856 on Build GRCh38
rs115424454 Chr.17: 8189834-8189834 on Build GRCh38
rs779261793 Chr.17: 8189772-8189772 on Build GRCh38
rs746018914 Chr.17: 8189706-8189706 on Build GRCh38
rs759408918 Chr.17: 8189813-8189813 on Build GRCh38
rs528851609 Chr.17: 8189818-8189818 on Build GRCh38
rs749428825 Chr.17: 8189690-8189690 on Build GRCh38
rs756250074 Chr.17: 8189713-8189715 on Build GRCh38
rs982019575 Chr.17: 8189691-8189691 on Build GRCh38
rs747455049 Chr.17: 8189784-8189784 on Build GRCh38
rs752429991 Chr.17: 8189816-8189816 on Build GRCh38
rs750619625 Chr.17: 8189820-8189820 on Build GRCh38
rs959307403 Chr.17: 8189838-8189838 on Build GRCh38
rs755540795 Chr.17: 8189832-8189832 on Build GRCh38
rs962873911 Chr.17: 8189799-8189799 on Build GRCh38
rs769552118 Chr.17: 8189744-8189744 on Build GRCh38
rs1010332391 Chr.17: 8189717-8189717 on Build GRCh38
rs777142396 Chr.17: 8189741-8189741 on Build GRCh38
rs562595644 Chr.17: 8189853-8189853 on Build GRCh38
rs757406881 Chr.17: 8189762-8189762 on Build GRCh38
rs937334521 Chr.17: 8189850-8189850 on Build GRCh38
rs780313893 Chr.17: 8189783-8189783 on Build GRCh38
rs751446913 Chr.17: 8189749-8189749 on Build GRCh38
rs561853521 Chr.17: 8189815-8189815 on Build GRCh38
rs1049980190 Chr.17: 8189792-8189792 on Build GRCh38
rs989136980 Chr.17: 8189849-8189849 on Build GRCh38
rs747541858 Chr.17: 8189732-8189732 on Build GRCh38
rs772452730 Chr.17: 8189711-8189711 on Build GRCh38
rs752549079 Chr.17: 8189758-8189758 on Build GRCh38
rs753966049 Chr.17: 8189748-8189748 on Build GRCh38
rs765064766 Chr.17: 8189858-8189858 on Build GRCh38
rs760558175 Chr.17: 8189817-8189817 on Build GRCh38
rs780985697 Chr.17: 8189672-8189672 on Build GRCh38
rs551741051 Chr.17: 8189870-8189870 on Build GRCh38
rs767230907 Chr.17: 8189756-8189756 on Build GRCh38
rs758700053 Chr.17: 8189821-8189821 on Build GRCh38
rs778798504 Chr.17: 8189696-8189696 on Build GRCh38
rs966367446 Chr.17: 8189868-8189868 on Build GRCh38
rs780227387 Chr.17: 8189827-8189827 on Build GRCh38
rs769302085 Chr.17: 8189788-8189788 on Build GRCh38
rs770384390 Chr.17: 8189805-8189805 on Build GRCh38

View Full Product Details