Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37004471-37004714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC8
Entrez Gene ID
11340
Gene Name
exosome component 8
Gene Aliases
CIP3, EAP2, OIP2, PCH1C, RRP43, Rrp43p, bA421P11.3, p9
UniGene
Hs.294041
Gene Chromosome Location
Chr.13: 37000373-37009614 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
469397 c.202G>C p.A68P Chr.13: 37004525-37004525 on Build GRCh38
1562263 c.234C>T p.Y78Y Chr.13: 37004557-37004557 on Build GRCh38
6384455 c.217G>T p.A73S Chr.13: 37004540-37004540 on Build GRCh38
223639 c.220C>T p.P74S Chr.13: 37004543-37004543 on Build GRCh38
5637362 c.232T>A p.Y78N Chr.13: 37004555-37004555 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753939023 Chr.13: 37004669-37004669 on Build GRCh38
rs746375170 Chr.13: 37004573-37004573 on Build GRCh38
rs149303311 Chr.13: 37004534-37004534 on Build GRCh38
rs376679078 Chr.13: 37004605-37004605 on Build GRCh38
rs750340849 Chr.13: 37004513-37004513 on Build GRCh38
rs547107309 Chr.13: 37004574-37004574 on Build GRCh38
rs982964134 Chr.13: 37004617-37004617 on Build GRCh38
rs767250476 Chr.13: 37004507-37004507 on Build GRCh38
rs748742813 Chr.13: 37004583-37004583 on Build GRCh38
rs774017612 Chr.13: 37004597-37004597 on Build GRCh38
rs756052584 Chr.13: 37004522-37004522 on Build GRCh38
rs765390835 Chr.13: 37004523-37004523 on Build GRCh38
rs747216574 Chr.13: 37004557-37004557 on Build GRCh38
rs908689054 Chr.13: 37004637-37004637 on Build GRCh38
rs765433320 Chr.13: 37004673-37004673 on Build GRCh38
rs963059202 Chr.13: 37004524-37004524 on Build GRCh38
rs752887601 Chr.13: 37004531-37004531 on Build GRCh38
rs565214972 Chr.13: 37004577-37004577 on Build GRCh38
rs768181529 Chr.13: 37004596-37004596 on Build GRCh38
rs773423674 Chr.13: 37004545-37004545 on Build GRCh38
rs577835326 Chr.13: 37004594-37004594 on Build GRCh38
rs960936403 Chr.13: 37004602-37004602 on Build GRCh38
rs199679550 Chr.13: 37004539-37004539 on Build GRCh38
rs1035663362 Chr.13: 37004559-37004559 on Build GRCh38
rs916263378 Chr.13: 37004538-37004538 on Build GRCh38
rs781460199 Chr.13: 37004572-37004572 on Build GRCh38
rs947782197 Chr.13: 37004551-37004551 on Build GRCh38
rs778083665 Chr.13: 37004540-37004540 on Build GRCh38
rs757600036 Chr.13: 37004558-37004558 on Build GRCh38
rs1053368635 Chr.13: 37004685-37004685 on Build GRCh38

View Full Product Details