Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54940290-54940543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83B
Entrez Gene ID
222584
Gene Name
family with sequence similarity 83 member B
Gene Aliases
C6orf143
UniGene
Hs.152423
Gene Chromosome Location
Chr.6: 54846323-54945099 on Build GRCh38
Transcripts
NM_001010872.2 XM_006715022.3 XM_011514394.2 XM_011514395.2 XM_017010478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305817 c.1503A>C p.L501F Chr.6: 54940474-54940474 on Build GRCh38
6107390 c.1485G>T p.W495C Chr.6: 54940456-54940456 on Build GRCh38
210970 c.1537G>T p.G513* Chr.6: 54940508-54940508 on Build GRCh38
1621782 c.1393A>T p.R465W Chr.6: 54940364-54940364 on Build GRCh38
5808355 c.1434A>G p.Q478Q Chr.6: 54940405-54940405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770940709 Chr.6: 54940409-54940409 on Build GRCh38
rs774932199 Chr.6: 54940511-54940511 on Build GRCh38
rs373419492 Chr.6: 54940448-54940448 on Build GRCh38
rs766009606 Chr.6: 54940491-54940491 on Build GRCh38
rs367671422 Chr.6: 54940371-54940371 on Build GRCh38
rs754426016 Chr.6: 54940510-54940510 on Build GRCh38
rs766679622 Chr.6: 54940366-54940366 on Build GRCh38
rs893782748 Chr.6: 54940397-54940397 on Build GRCh38
rs746725945 Chr.6: 54940492-54940492 on Build GRCh38
rs995852319 Chr.6: 54940476-54940476 on Build GRCh38
rs138195667 Chr.6: 54940368-54940368 on Build GRCh38
rs768020820 Chr.6: 54940514-54940514 on Build GRCh38
rs750088415 Chr.6: 54940410-54940410 on Build GRCh38
rs764861449 Chr.6: 54940470-54940470 on Build GRCh38
rs776540555 Chr.6: 54940336-54940336 on Build GRCh38
rs375437689 Chr.6: 54940418-54940418 on Build GRCh38
rs749436785 Chr.6: 54940415-54940416 on Build GRCh38
rs778923838 Chr.6: 54940395-54940395 on Build GRCh38
rs367910496 Chr.6: 54940403-54940403 on Build GRCh38
rs945654716 Chr.6: 54940358-54940358 on Build GRCh38
rs149380682 Chr.6: 54940498-54940498 on Build GRCh38
rs761941208 Chr.6: 54940337-54940337 on Build GRCh38
rs201956396 Chr.6: 54940329-54940329 on Build GRCh38
rs148485145 Chr.6: 54940472-54940476 on Build GRCh38
rs561363029 Chr.6: 54940468-54940468 on Build GRCh38
rs768749527 Chr.6: 54940414-54940414 on Build GRCh38
rs775741755 Chr.6: 54940321-54940321 on Build GRCh38
rs769893635 Chr.6: 54940421-54940421 on Build GRCh38
rs369789741 Chr.6: 54940434-54940434 on Build GRCh38
rs755694372 Chr.6: 54940378-54940378 on Build GRCh38
rs749453308 Chr.6: 54940507-54940507 on Build GRCh38
rs375679054 Chr.6: 54940361-54940361 on Build GRCh38
rs777546716 Chr.6: 54940380-54940380 on Build GRCh38
rs776374003 Chr.6: 54940519-54940519 on Build GRCh38
rs761534241 Chr.6: 54940521-54940521 on Build GRCh38
rs368876321 Chr.6: 54940497-54940497 on Build GRCh38
rs781353175 Chr.6: 54940496-54940496 on Build GRCh38
rs201372672 Chr.6: 54940398-54940398 on Build GRCh38
rs759749145 Chr.6: 54940512-54940512 on Build GRCh38
rs147775708 Chr.6: 54940317-54940317 on Build GRCh38
rs760828107 Chr.6: 54940328-54940328 on Build GRCh38
rs1029940353 Chr.6: 54940432-54940432 on Build GRCh38
rs138570262 Chr.6: 54940494-54940494 on Build GRCh38
rs141100648 Chr.6: 54940318-54940318 on Build GRCh38
rs140860872 Chr.6: 54940417-54940417 on Build GRCh38
rs371794676 Chr.6: 54940382-54940382 on Build GRCh38
rs750001073 Chr.6: 54940471-54940471 on Build GRCh38
rs376789656 Chr.6: 54940394-54940394 on Build GRCh38
rs866073707 Chr.6: 54940504-54940504 on Build GRCh38
rs752080297 Chr.6: 54940385-54940385 on Build GRCh38
rs147022305 Chr.6: 54940362-54940362 on Build GRCh38
rs763592072 Chr.6: 54940465-54940465 on Build GRCh38
rs115872183 Chr.6: 54940481-54940481 on Build GRCh38
rs553145456 Chr.6: 54940310-54940310 on Build GRCh38
rs748850090 Chr.6: 54940315-54940315 on Build GRCh38
rs573151626 Chr.6: 54940311-54940311 on Build GRCh38
rs866880366 Chr.6: 54940312-54940312 on Build GRCh38
rs771764348 Chr.6: 54940320-54940320 on Build GRCh38
rs376811005 Chr.6: 54940449-54940449 on Build GRCh38
rs773665194 Chr.6: 54940430-54940430 on Build GRCh38

View Full Product Details