Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 119121038-119121263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
UniGene
Hs.181042
Gene Chromosome Location
Chr.5: 119071002-119250170 on Build GRCh38
Transcripts
NM_001290321.2 NM_001290322.2 NM_005509.5 XM_005271909.4 XM_005271910.4 XM_005271912.2 XM_011543212.2 XM_011543213.2 XM_011543214.2 XM_011543215.2 XM_017009141.1 XM_017009142.1 XM_017009143.1 XM_017009144.1 XM_017009145.1 XM_017009146.1 XM_017009147.1 XM_017009148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950446 c.1088T>G p.I363S Chr.5: 119121125-119121125 on Build GRCh38
7123092 c.1069_1101del33 p.F357_T367del11 Chr.5: 119121106-119121138 on Build GRCh38
4950445 c.1088T>G p.I363S Chr.5: 119121125-119121125 on Build GRCh38
1580780 c.1090A>G p.N364D Chr.5: 119121127-119121127 on Build GRCh38
1250286 c.1070T>A p.F357Y Chr.5: 119121107-119121107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766703061 Chr.5: 119121104-119121104 on Build GRCh38
rs941097501 Chr.5: 119121202-119121202 on Build GRCh38
rs960938188 Chr.5: 119121110-119121110 on Build GRCh38
rs774225987 Chr.5: 119121094-119121094 on Build GRCh38
rs367905216 Chr.5: 119121090-119121090 on Build GRCh38
rs779444486 Chr.5: 119121072-119121072 on Build GRCh38
rs148466434 Chr.5: 119121076-119121076 on Build GRCh38
rs764584706 Chr.5: 119121185-119121185 on Build GRCh38
rs771327879 Chr.5: 119121173-119121173 on Build GRCh38
rs753279192 Chr.5: 119121130-119121130 on Build GRCh38
rs552479832 Chr.5: 119121223-119121223 on Build GRCh38
rs777013804 Chr.5: 119121174-119121174 on Build GRCh38
rs756974807 Chr.5: 119121136-119121136 on Build GRCh38
rs754011601 Chr.5: 119121109-119121109 on Build GRCh38
rs371356301 Chr.5: 119121165-119121165 on Build GRCh38
rs765180977 Chr.5: 119121128-119121128 on Build GRCh38
rs751964815 Chr.5: 119121188-119121188 on Build GRCh38
rs774350695 Chr.5: 119121167-119121167 on Build GRCh38
rs937750197 Chr.5: 119121059-119121059 on Build GRCh38
rs562921376 Chr.5: 119121092-119121092 on Build GRCh38
rs762757255 Chr.5: 119121091-119121091 on Build GRCh38
rs765365631 Chr.5: 119121182-119121182 on Build GRCh38
rs761668941 Chr.5: 119121170-119121170 on Build GRCh38
rs761033172 Chr.5: 119121103-119121103 on Build GRCh38
rs145511612 Chr.5: 119121073-119121073 on Build GRCh38
rs755085610 Chr.5: 119121121-119121121 on Build GRCh38
rs763574063 Chr.5: 119121184-119121184 on Build GRCh38
rs374647823 Chr.5: 119121175-119121175 on Build GRCh38
rs757598468 Chr.5: 119121191-119121191 on Build GRCh38
rs1010213539 Chr.5: 119121212-119121212 on Build GRCh38
rs968513024 Chr.5: 119121159-119121159 on Build GRCh38
rs769661786 Chr.5: 119121089-119121089 on Build GRCh38
rs909578266 Chr.5: 119121178-119121178 on Build GRCh38
rs896979283 Chr.5: 119121211-119121211 on Build GRCh38
rs891903939 Chr.5: 119121209-119121209 on Build GRCh38
rs745381631 Chr.5: 119121088-119121088 on Build GRCh38
rs779360982 Chr.5: 119121157-119121157 on Build GRCh38
rs746670060 Chr.5: 119121077-119121077 on Build GRCh38
rs776185916 Chr.5: 119121085-119121085 on Build GRCh38
rs749259806 Chr.5: 119121163-119121163 on Build GRCh38
rs752663003 Chr.5: 119121183-119121183 on Build GRCh38
rs769372230 Chr.5: 119121156-119121156 on Build GRCh38
rs563241866 Chr.5: 119121180-119121180 on Build GRCh38
rs780795841 Chr.5: 119121143-119121143 on Build GRCh38
rs532482648 Chr.5: 119121206-119121206 on Build GRCh38
rs778214012 Chr.5: 119121132-119121132 on Build GRCh38
rs373648460 Chr.5: 119121151-119121151 on Build GRCh38

View Full Product Details