Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49643453-49643726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT5
Entrez Gene ID
9334
Gene Name
beta-1,4-galactosyltransferase 5
Gene Aliases
B4Gal-T5, BETA4-GALT-IV, beta4Gal-T5, beta4GalT-V, gt-V
UniGene
Hs.370487
Gene Chromosome Location
Chr.20: 49632945-49713884 on Build GRCh38
Transcripts
NM_004776.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4496630 c.479C>T p.P160L Chr.20: 49643536-49643536 on Build GRCh38
1483736 c.415G>C p.E139Q Chr.20: 49643600-49643600 on Build GRCh38
4493772 c.420C>T p.L140L Chr.20: 49643595-49643595 on Build GRCh38
2762256 c.467C>T p.S156F Chr.20: 49643548-49643548 on Build GRCh38
4846979 c.420C>G p.L140L Chr.20: 49643595-49643595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367589949 Chr.20: 49643563-49643563 on Build GRCh38
rs570510982 Chr.20: 49643488-49643488 on Build GRCh38
rs756374441 Chr.20: 49643658-49643658 on Build GRCh38
rs772951768 Chr.20: 49643595-49643595 on Build GRCh38
rs752067075 Chr.20: 49643502-49643502 on Build GRCh38
rs200335651 Chr.20: 49643642-49643642 on Build GRCh38
rs760073543 Chr.20: 49643641-49643641 on Build GRCh38
rs770665322 Chr.20: 49643520-49643520 on Build GRCh38
rs776281104 Chr.20: 49643583-49643583 on Build GRCh38
rs140018307 Chr.20: 49643620-49643620 on Build GRCh38
rs745636592 Chr.20: 49643548-49643548 on Build GRCh38
rs368497312 Chr.20: 49643572-49643572 on Build GRCh38
rs746894463 Chr.20: 49643576-49643576 on Build GRCh38
rs953856378 Chr.20: 49643683-49643683 on Build GRCh38
rs369840065 Chr.20: 49643662-49643662 on Build GRCh38
rs369348234 Chr.20: 49643696-49643696 on Build GRCh38
rs752741936 Chr.20: 49643643-49643643 on Build GRCh38
rs1018864768 Chr.20: 49643479-49643479 on Build GRCh38
rs780902442 Chr.20: 49643698-49643698 on Build GRCh38
rs766817510 Chr.20: 49643494-49643494 on Build GRCh38
rs769549483 Chr.20: 49643590-49643590 on Build GRCh38
rs768111785 Chr.20: 49643582-49643582 on Build GRCh38
rs568690842 Chr.20: 49643538-49643538 on Build GRCh38
rs773961414 Chr.20: 49643634-49643634 on Build GRCh38
rs759023992 Chr.20: 49643483-49643483 on Build GRCh38
rs750789772 Chr.20: 49643482-49643482 on Build GRCh38
rs373671697 Chr.20: 49643540-49643540 on Build GRCh38
rs6122802 Chr.20: 49643666-49643666 on Build GRCh38
rs774875274 Chr.20: 49643696-49643696 on Build GRCh38
rs1010228870 Chr.20: 49643587-49643587 on Build GRCh38
rs371958533 Chr.20: 49643593-49643593 on Build GRCh38
rs746989324 Chr.20: 49643679-49643679 on Build GRCh38
rs183754228 Chr.20: 49643663-49643663 on Build GRCh38
rs775319181 Chr.20: 49643574-49643574 on Build GRCh38
rs537895750 Chr.20: 49643505-49643505 on Build GRCh38
rs1008794521 Chr.20: 49643478-49643478 on Build GRCh38
rs779888829 Chr.20: 49643673-49643673 on Build GRCh38
rs146048963 Chr.20: 49643568-49643568 on Build GRCh38
rs761601631 Chr.20: 49643588-49643588 on Build GRCh38
rs766700526 Chr.20: 49643628-49643628 on Build GRCh38
rs550435557 Chr.20: 49643533-49643533 on Build GRCh38
rs752019385 Chr.20: 49643630-49643630 on Build GRCh38
rs879211591 Chr.20: 49643515-49643515 on Build GRCh38
rs755019668 Chr.20: 49643514-49643514 on Build GRCh38
rs769703631 Chr.20: 49643484-49643484 on Build GRCh38
rs748425737 Chr.20: 49643537-49643537 on Build GRCh38

View Full Product Details