Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26943328-26943559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA5
Entrez Gene ID
2558
Gene Name
gamma-aminobutyric acid type A receptor alpha5 subunit
Gene Aliases
-
UniGene
Hs.24969
Gene Chromosome Location
Chr.15: 26866719-26949210 on Build GRCh38
Transcripts
NM_000810.3 NM_001165037.1 XM_005268258.2 XM_006720459.2 XM_017022055.1 XM_017022056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726003 c.1029C>G p.Y343* Chr.15: 26943366-26943366 on Build GRCh38
1252956 c.1060delA p.A356fs*15 Chr.15: 26943397-26943397 on Build GRCh38
700318 c.1043G>A p.G348D Chr.15: 26943380-26943380 on Build GRCh38
1372198 c.1058G>A p.G353D Chr.15: 26943395-26943395 on Build GRCh38
6360787 c.1045T>C p.W349R Chr.15: 26943382-26943382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774379564 Chr.15: 26943440-26943440 on Build GRCh38
rs75690401 Chr.15: 26943357-26943357 on Build GRCh38
rs192324022 Chr.15: 26943525-26943525 on Build GRCh38
rs569156389 Chr.15: 26943506-26943506 on Build GRCh38
rs758283106 Chr.15: 26943397-26943397 on Build GRCh38
rs866250154 Chr.15: 26943353-26943353 on Build GRCh38
rs771713182 Chr.15: 26943479-26943479 on Build GRCh38
rs772245945 Chr.15: 26943472-26943472 on Build GRCh38
rs770196016 Chr.15: 26943415-26943415 on Build GRCh38
rs1050457253 Chr.15: 26943466-26943466 on Build GRCh38
rs749578040 Chr.15: 26943435-26943435 on Build GRCh38
rs762019527 Chr.15: 26943465-26943465 on Build GRCh38
rs775948242 Chr.15: 26943433-26943433 on Build GRCh38
rs773088112 Chr.15: 26943533-26943533 on Build GRCh38
rs184713635 Chr.15: 26943526-26943526 on Build GRCh38
rs548961867 Chr.15: 26943436-26943436 on Build GRCh38
rs902407170 Chr.15: 26943488-26943488 on Build GRCh38
rs946376818 Chr.15: 26943461-26943461 on Build GRCh38
rs1043400018 Chr.15: 26943485-26943485 on Build GRCh38
rs746347028 Chr.15: 26943396-26943396 on Build GRCh38
rs41317330 Chr.15: 26943354-26943354 on Build GRCh38

View Full Product Details