Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15272416-15272643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4222448 c.251T>G p.F84C Chr.1: 15272480-15272480 on Build GRCh38
4951865 c.229A>G p.I77V Chr.1: 15272458-15272458 on Build GRCh38
4647355 c.195C>T p.N65N Chr.1: 15272424-15272424 on Build GRCh38
5035892 c.292C>G p.P98A Chr.1: 15272521-15272521 on Build GRCh38
6400721 c.217G>A p.A73T Chr.1: 15272446-15272446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75432354 Chr.1: 15272529-15272529 on Build GRCh38
rs576057104 Chr.1: 15272484-15272484 on Build GRCh38
rs190738579 Chr.1: 15272537-15272537 on Build GRCh38
rs894026757 Chr.1: 15272538-15272538 on Build GRCh38
rs778304960 Chr.1: 15272567-15272567 on Build GRCh38
rs765416342 Chr.1: 15272443-15272443 on Build GRCh38
rs750593539 Chr.1: 15272495-15272501 on Build GRCh38
rs763015518 Chr.1: 15272488-15272488 on Build GRCh38
rs6679905 Chr.1: 15272525-15272525 on Build GRCh38
rs185146952 Chr.1: 15272532-15272532 on Build GRCh38
rs908372567 Chr.1: 15272444-15272444 on Build GRCh38
rs531379972 Chr.1: 15272603-15272603 on Build GRCh38
rs538645934 Chr.1: 15272502-15272502 on Build GRCh38
rs542775831 Chr.1: 15272581-15272581 on Build GRCh38
rs779616398 Chr.1: 15272536-15272536 on Build GRCh38
rs751376805 Chr.1: 15272523-15272523 on Build GRCh38
rs769245323 Chr.1: 15272582-15272582 on Build GRCh38
rs914976620 Chr.1: 15272620-15272620 on Build GRCh38
rs374554789 Chr.1: 15272608-15272608 on Build GRCh38
rs775970258 Chr.1: 15272556-15272556 on Build GRCh38
rs1003753218 Chr.1: 15272586-15272586 on Build GRCh38
rs763931431 Chr.1: 15272499-15272499 on Build GRCh38
rs995086006 Chr.1: 15272524-15272524 on Build GRCh38
rs56987602 Chr.1: 15272573-15272573 on Build GRCh38
rs767070635 Chr.1: 15272528-15272528 on Build GRCh38
rs562545155 Chr.1: 15272589-15272589 on Build GRCh38
rs961864989 Chr.1: 15272521-15272521 on Build GRCh38
rs764496026 Chr.1: 15272584-15272584 on Build GRCh38

View Full Product Details