Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 52752019-52752258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCC
Entrez Gene ID
1630
Gene Name
DCC netrin 1 receptor
Gene Aliases
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1
UniGene
Hs.162025
Gene Chromosome Location
Chr.18: 52340172-53535903 on Build GRCh38
Transcripts
NM_005215.3 XM_011525843.1 XM_017025568.1 XM_017025569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7208683 c.161G>T p.G54V Chr.18: 52752123-52752123 on Build GRCh38
260366 c.140C>A p.S47Y Chr.18: 52752102-52752102 on Build GRCh38
5056069 c.148G>A p.V50I Chr.18: 52752110-52752110 on Build GRCh38
4676411 c.209C>T p.P70L Chr.18: 52752171-52752171 on Build GRCh38
988919 c.136C>T p.P46S Chr.18: 52752098-52752098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038717902 Chr.18: 52752172-52752172 on Build GRCh38
rs199598105 Chr.18: 52752181-52752181 on Build GRCh38
rs777555256 Chr.18: 52752109-52752109 on Build GRCh38
rs112141078 Chr.18: 52752193-52752193 on Build GRCh38
rs781178843 Chr.18: 52752213-52752213 on Build GRCh38
rs1040684774 Chr.18: 52752075-52752075 on Build GRCh38
rs376690246 Chr.18: 52752082-52752082 on Build GRCh38
rs893585925 Chr.18: 52752173-52752173 on Build GRCh38
rs146575994 Chr.18: 52752162-52752162 on Build GRCh38
rs990772793 Chr.18: 52752078-52752078 on Build GRCh38
rs369755020 Chr.18: 52752085-52752085 on Build GRCh38
rs775883937 Chr.18: 52752116-52752116 on Build GRCh38
rs769007732 Chr.18: 52752230-52752230 on Build GRCh38
rs752208238 Chr.18: 52752165-52752165 on Build GRCh38
rs962808767 Chr.18: 52752083-52752083 on Build GRCh38
rs141272344 Chr.18: 52752180-52752180 on Build GRCh38
rs1042628440 Chr.18: 52752209-52752209 on Build GRCh38
rs764721371 Chr.18: 52752163-52752163 on Build GRCh38
rs911570615 Chr.18: 52752133-52752133 on Build GRCh38
rs1033102782 Chr.18: 52752212-52752212 on Build GRCh38
rs768831554 Chr.18: 52752120-52752120 on Build GRCh38
rs749521328 Chr.18: 52752220-52752220 on Build GRCh38
rs774517981 Chr.18: 52752231-52752231 on Build GRCh38
rs145690431 Chr.18: 52752126-52752126 on Build GRCh38
rs761298636 Chr.18: 52752047-52752047 on Build GRCh38
rs148342408 Chr.18: 52752196-52752196 on Build GRCh38
rs750760650 Chr.18: 52752063-52752063 on Build GRCh38
rs767923423 Chr.18: 52752047-52752047 on Build GRCh38
rs200975371 Chr.18: 52752091-52752091 on Build GRCh38
rs762319358 Chr.18: 52752137-52752137 on Build GRCh38
rs766772715 Chr.18: 52752157-52752157 on Build GRCh38
rs756966505 Chr.18: 52752202-52752202 on Build GRCh38
rs749558240 Chr.18: 52752119-52752119 on Build GRCh38
rs779596593 Chr.18: 52752077-52752077 on Build GRCh38
rs190581937 Chr.18: 52752113-52752113 on Build GRCh38
rs552730020 Chr.18: 52752195-52752195 on Build GRCh38
rs144331156 Chr.18: 52752158-52752158 on Build GRCh38
rs778444286 Chr.18: 52752086-52752086 on Build GRCh38
rs202005334 Chr.18: 52752084-52752084 on Build GRCh38
rs755736951 Chr.18: 52752069-52752069 on Build GRCh38
rs760978364 Chr.18: 52752150-52752150 on Build GRCh38
rs558026226 Chr.18: 52752149-52752149 on Build GRCh38
rs767925738 Chr.18: 52752148-52752148 on Build GRCh38
rs774643012 Chr.18: 52752121-52752121 on Build GRCh38
rs148156622 Chr.18: 52752110-52752110 on Build GRCh38
rs758335110 Chr.18: 52752095-52752095 on Build GRCh38

View Full Product Details