Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51346931-51347172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFB
Entrez Gene ID
2109
Gene Name
electron transfer flavoprotein beta subunit
Gene Aliases
FP585, MADD
UniGene
Hs.348531
Gene Chromosome Location
Chr.19: 51345155-51366418 on Build GRCh38
Transcripts
NM_001014763.1 NM_001985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6257586 c.564C>A p.N188K Chr.19: 51346933-51346933 on Build GRCh38
1734940 c.721G>A p.A241T Chr.19: 51347049-51347049 on Build GRCh38
6285427 c.507C>G p.G169G Chr.19: 51346990-51346990 on Build GRCh38
3537591 c.454C>T p.Q152* Chr.19: 51347043-51347043 on Build GRCh38
5095820 c.809C>T p.A270V Chr.19: 51346961-51346961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766809815 Chr.19: 51347073-51347073 on Build GRCh38
rs8112687 Chr.19: 51347148-51347148 on Build GRCh38
rs148443220 Chr.19: 51346956-51346956 on Build GRCh38
rs141917423 Chr.19: 51346976-51346976 on Build GRCh38
rs755291156 Chr.19: 51347078-51347078 on Build GRCh38
rs753045611 Chr.19: 51347082-51347082 on Build GRCh38
rs765846018 Chr.19: 51347026-51347026 on Build GRCh38
rs984189393 Chr.19: 51347112-51347112 on Build GRCh38
rs753381231 Chr.19: 51347054-51347054 on Build GRCh38
rs908909594 Chr.19: 51347128-51347128 on Build GRCh38
rs997125626 Chr.19: 51347067-51347067 on Build GRCh38
rs749396395 Chr.19: 51347022-51347022 on Build GRCh38
rs903169408 Chr.19: 51347025-51347025 on Build GRCh38
rs767451791 Chr.19: 51347040-51347040 on Build GRCh38
rs756770328 Chr.19: 51347072-51347072 on Build GRCh38
rs543044180 Chr.19: 51347090-51347090 on Build GRCh38
rs763680358 Chr.19: 51346979-51346979 on Build GRCh38
rs140614695 Chr.19: 51346998-51346998 on Build GRCh38
rs972038317 Chr.19: 51347145-51347145 on Build GRCh38
rs1011643509 Chr.19: 51347005-51347005 on Build GRCh38
rs575702007 Chr.19: 51347086-51347086 on Build GRCh38
rs778952850 Chr.19: 51347107-51347107 on Build GRCh38
rs866913724 Chr.19: 51347085-51347085 on Build GRCh38
rs1036677852 Chr.19: 51346970-51346970 on Build GRCh38
rs771139953 Chr.19: 51347104-51347104 on Build GRCh38
rs561512829 Chr.19: 51347110-51347110 on Build GRCh38
rs1130426 Chr.19: 51347036-51347036 on Build GRCh38
rs777916916 Chr.19: 51347088-51347088 on Build GRCh38
rs770890099 Chr.19: 51347029-51347029 on Build GRCh38
rs760351587 Chr.19: 51347049-51347049 on Build GRCh38
rs1023051509 Chr.19: 51347074-51347074 on Build GRCh38
rs781516814 Chr.19: 51347081-51347081 on Build GRCh38
rs148045813 Chr.19: 51347007-51347007 on Build GRCh38
rs150636733 Chr.19: 51346999-51346999 on Build GRCh38
rs144640661 Chr.19: 51347050-51347050 on Build GRCh38
rs749331575 Chr.19: 51347094-51347094 on Build GRCh38
rs773313415 Chr.19: 51347016-51347016 on Build GRCh38
rs774444434 Chr.19: 51347031-51347031 on Build GRCh38
rs74735908 Chr.19: 51347045-51347045 on Build GRCh38
rs529633264 Chr.19: 51347144-51347144 on Build GRCh38
rs780515260 Chr.19: 51347047-51347047 on Build GRCh38
rs908624750 Chr.19: 51347123-51347123 on Build GRCh38
rs755539437 Chr.19: 51346977-51346977 on Build GRCh38
rs933129897 Chr.19: 51346978-51346978 on Build GRCh38
rs370000302 Chr.19: 51347108-51347108 on Build GRCh38
rs758905314 Chr.19: 51346987-51346987 on Build GRCh38
rs752095831 Chr.19: 51347076-51347076 on Build GRCh38
rs753337486 Chr.19: 51346981-51346981 on Build GRCh38
rs141774128 Chr.19: 51347012-51347012 on Build GRCh38
rs104894677 Chr.19: 51347006-51347006 on Build GRCh38
rs182596750 Chr.19: 51347149-51347149 on Build GRCh38

View Full Product Details