Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66493094-66493367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEAN1
Entrez Gene ID
146227
Gene Name
brain expressed, associated with NEDD4, 1
Gene Aliases
BEAN, SCA31
UniGene
Hs.740218
Gene Chromosome Location
Chr.16: 66427297-66495288 on Build GRCh38
Transcripts
NM_001197224.3 NM_001197225.3

SNP Detail

SNP ID Chromosome Location
rs954537868 Chr.16: 66493186-66493186 on Build GRCh38
rs915400941 Chr.16: 66493121-66493121 on Build GRCh38
rs1037534276 Chr.16: 66493173-66493173 on Build GRCh38
rs967433130 Chr.16: 66493242-66493242 on Build GRCh38
rs781588018 Chr.16: 66493262-66493262 on Build GRCh38
rs776824964 Chr.16: 66493213-66493213 on Build GRCh38
rs536110926 Chr.16: 66493151-66493151 on Build GRCh38
rs138344650 Chr.16: 66493143-66493143 on Build GRCh38
rs897567506 Chr.16: 66493176-66493176 on Build GRCh38
rs115852696 Chr.16: 66493162-66493162 on Build GRCh38
rs990308735 Chr.16: 66493231-66493231 on Build GRCh38
rs866965717 Chr.16: 66493184-66493184 on Build GRCh38
rs1029972596 Chr.16: 66493185-66493185 on Build GRCh38
rs993233050 Chr.16: 66493181-66493181 on Build GRCh38
rs981093840 Chr.16: 66493281-66493281 on Build GRCh38
rs754537210 Chr.16: 66493130-66493130 on Build GRCh38
rs946911210 Chr.16: 66493337-66493337 on Build GRCh38
rs527873790 Chr.16: 66493165-66493165 on Build GRCh38
rs117499742 Chr.16: 66493333-66493333 on Build GRCh38
rs567787747 Chr.16: 66493237-66493237 on Build GRCh38
rs1013284142 Chr.16: 66493224-66493224 on Build GRCh38
rs1004286389 Chr.16: 66493172-66493172 on Build GRCh38
rs762955232 Chr.16: 66493332-66493332 on Build GRCh38
rs764597671 Chr.16: 66493247-66493247 on Build GRCh38
rs977516969 Chr.16: 66493254-66493254 on Build GRCh38
rs139137875 Chr.16: 66493328-66493328 on Build GRCh38
rs771159228 Chr.16: 66493202-66493202 on Build GRCh38
rs938684168 Chr.16: 66493280-66493280 on Build GRCh38
rs58468753 Chr.16: 66493289-66493289 on Build GRCh38
rs9928279 Chr.16: 66493196-66493196 on Build GRCh38
rs747652408 Chr.16: 66493161-66493161 on Build GRCh38
rs149611294 Chr.16: 66493150-66493150 on Build GRCh38
rs964328105 Chr.16: 66493229-66493229 on Build GRCh38
rs894270005 Chr.16: 66493163-66493163 on Build GRCh38
rs747737058 Chr.16: 66493164-66493164 on Build GRCh38
rs865856531 Chr.16: 66493136-66493136 on Build GRCh38
rs146861397 Chr.16: 66493290-66493290 on Build GRCh38

View Full Product Details