Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 139015975-139016200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOCT
Entrez Gene ID
25819
Gene Name
nocturnin
Gene Aliases
CCR4L, CCRN4L, Ccr4c, NOC
UniGene
Hs.639842
Gene Chromosome Location
Chr.4: 139015759-139045939 on Build GRCh38
Transcripts
NM_012118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4138049 c.119G>T p.R40M Chr.4: 139016100-139016100 on Build GRCh38
6203540 c.19C>A p.R7R Chr.4: 139016000-139016000 on Build GRCh38
6033994 c.128delC p.R45fs*38 Chr.4: 139016109-139016109 on Build GRCh38
5662864 c.44G>T p.R15M Chr.4: 139016025-139016025 on Build GRCh38
4657591 c.136C>T p.L46L Chr.4: 139016117-139016117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898848357 Chr.4: 139016180-139016180 on Build GRCh38
rs186938335 Chr.4: 139016080-139016080 on Build GRCh38
rs750292631 Chr.4: 139016154-139016154 on Build GRCh38
rs183350335 Chr.4: 139016013-139016013 on Build GRCh38
rs775735506 Chr.4: 139016074-139016074 on Build GRCh38
rs796396566 Chr.4: 139016137-139016137 on Build GRCh38
rs548389864 Chr.4: 139016034-139016034 on Build GRCh38
rs943299857 Chr.4: 139016166-139016166 on Build GRCh38
rs977202712 Chr.4: 139016114-139016114 on Build GRCh38
rs780645746 Chr.4: 139016151-139016151 on Build GRCh38
rs916190552 Chr.4: 139016159-139016159 on Build GRCh38
rs545407418 Chr.4: 139016067-139016067 on Build GRCh38
rs537375822 Chr.4: 139016102-139016102 on Build GRCh38
rs1038692420 Chr.4: 139016171-139016171 on Build GRCh38
rs917477967 Chr.4: 139016070-139016070 on Build GRCh38
rs1026463178 Chr.4: 139016059-139016059 on Build GRCh38
rs1356461 Chr.4: 139016063-139016063 on Build GRCh38
rs1009473416 Chr.4: 139015997-139015997 on Build GRCh38
rs759598367 Chr.4: 139016057-139016057 on Build GRCh38
rs936098728 Chr.4: 139016109-139016109 on Build GRCh38
rs561885304 Chr.4: 139016001-139016001 on Build GRCh38
rs945167132 Chr.4: 139016156-139016156 on Build GRCh38
rs1017693611 Chr.4: 139016026-139016026 on Build GRCh38
rs890396248 Chr.4: 139016011-139016011 on Build GRCh38
rs1051600217 Chr.4: 139016157-139016157 on Build GRCh38
rs890342129 Chr.4: 139016164-139016164 on Build GRCh38
rs995033462 Chr.4: 139016046-139016046 on Build GRCh38
rs569819495 Chr.4: 139016091-139016091 on Build GRCh38
rs757712906 Chr.4: 139016124-139016132 on Build GRCh38
rs552158252 Chr.4: 139016117-139016117 on Build GRCh38
rs879589061 Chr.4: 139016061-139016061 on Build GRCh38
rs570473272 Chr.4: 139016160-139016160 on Build GRCh38
rs992972100 Chr.4: 139016069-139016069 on Build GRCh38
rs534646574 Chr.4: 139016165-139016165 on Build GRCh38
rs1053637878 Chr.4: 139016111-139016111 on Build GRCh38

View Full Product Details