Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11964623-11964848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK3
Entrez Gene ID
27122
Gene Name
dickkopf WNT signaling pathway inhibitor 3
Gene Aliases
REIC, RIG
UniGene
Hs.292156
Gene Chromosome Location
Chr.11: 11962996-12009370 on Build GRCh38
Transcripts
NM_001018057.1 NM_013253.4 NM_015881.5 XM_006718178.2 XM_017017553.1 XM_017017554.1 XM_017017555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475118 c.839_840insT p.V281fs*13 Chr.11: 11964677-11964678 on Build GRCh38
5477452 c.872G>A p.S291N Chr.11: 11964645-11964645 on Build GRCh38
3444229 c.875G>A p.R292H Chr.11: 11964642-11964642 on Build GRCh38
7308623 c.857C>T p.P286L Chr.11: 11964660-11964660 on Build GRCh38
4617084 c.848T>A p.V283E Chr.11: 11964669-11964669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767584922 Chr.11: 11964725-11964725 on Build GRCh38
rs775082546 Chr.11: 11964728-11964728 on Build GRCh38
rs547603749 Chr.11: 11964733-11964733 on Build GRCh38
rs372860129 Chr.11: 11964721-11964721 on Build GRCh38
rs1016525719 Chr.11: 11964675-11964675 on Build GRCh38
rs868450387 Chr.11: 11964676-11964676 on Build GRCh38
rs373077042 Chr.11: 11964669-11964669 on Build GRCh38
rs185983490 Chr.11: 11964795-11964795 on Build GRCh38
rs780059527 Chr.11: 11964691-11964691 on Build GRCh38
rs553951047 Chr.11: 11964740-11964740 on Build GRCh38
rs770613373 Chr.11: 11964720-11964720 on Build GRCh38
rs924231086 Chr.11: 11964803-11964803 on Build GRCh38
rs969675561 Chr.11: 11964693-11964693 on Build GRCh38
rs752126438 Chr.11: 11964774-11964774 on Build GRCh38
rs758840695 Chr.11: 11964759-11964759 on Build GRCh38
rs779382266 Chr.11: 11964684-11964684 on Build GRCh38
rs756973312 Chr.11: 11964750-11964750 on Build GRCh38
rs532056198 Chr.11: 11964657-11964657 on Build GRCh38
rs146019449 Chr.11: 11964668-11964668 on Build GRCh38
rs891366151 Chr.11: 11964735-11964735 on Build GRCh38
rs776826425 Chr.11: 11964699-11964699 on Build GRCh38
rs563274961 Chr.11: 11964647-11964647 on Build GRCh38
rs746329146 Chr.11: 11964686-11964686 on Build GRCh38
rs544051087 Chr.11: 11964759-11964760 on Build GRCh38
rs765478359 Chr.11: 11964751-11964751 on Build GRCh38
rs774144698 Chr.11: 11964653-11964653 on Build GRCh38
rs565471244 Chr.11: 11964713-11964713 on Build GRCh38
rs1054771773 Chr.11: 11964818-11964818 on Build GRCh38
rs995968624 Chr.11: 11964756-11964756 on Build GRCh38
rs545673121 Chr.11: 11964659-11964659 on Build GRCh38
rs142651172 Chr.11: 11964666-11964666 on Build GRCh38
rs750821593 Chr.11: 11964752-11964752 on Build GRCh38
rs762657292 Chr.11: 11964649-11964649 on Build GRCh38
rs992111827 Chr.11: 11964779-11964779 on Build GRCh38
rs373767107 Chr.11: 11964652-11964652 on Build GRCh38
rs933759425 Chr.11: 11964769-11964769 on Build GRCh38
rs934349235 Chr.11: 11964805-11964805 on Build GRCh38
rs768797263 Chr.11: 11964698-11964698 on Build GRCh38
rs370166416 Chr.11: 11964739-11964739 on Build GRCh38
rs780540189 Chr.11: 11964764-11964764 on Build GRCh38
rs758974264 Chr.11: 11964687-11964687 on Build GRCh38
rs755778952 Chr.11: 11964660-11964660 on Build GRCh38
rs760268191 Chr.11: 11964734-11964734 on Build GRCh38
rs747101046 Chr.11: 11964692-11964692 on Build GRCh38

View Full Product Details