Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22006114-22006341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2B
Entrez Gene ID
1030
Gene Name
cyclin dependent kinase inhibitor 2B
Gene Aliases
CDK4I, INK4B, MTS2, P15, TP15, p15INK4b
UniGene
Hs.72901
Gene Chromosome Location
Chr.9: 22002903-22009313 on Build GRCh38
Transcripts
NM_004936.3 NM_078487.2 NR_003529.3 NR_047532.1 NR_047533.1 NR_047534.1 NR_047535.1 NR_047536.1 NR_047537.1 NR_047538.1 NR_047539.1 NR_047540.1 NR_047541.1 NR_047542.1 NR_047543.1 NR_120536.1

Gene Symbol
CDKN2B-AS1
Entrez Gene ID
100048912
Gene Name
CDKN2B antisense RNA 1
Gene Aliases
ANRIL, CDKN2B-AS, CDKN2BAS, NCRNA00089, PCAT12, p15AS
UniGene
Hs.493614
Gene Chromosome Location
Chr.9: 21994791-22121097 on Build GRCh38
Transcripts
NM_004936.3 NM_078487.2 NR_003529.3 NR_047532.1 NR_047533.1 NR_047534.1 NR_047535.1 NR_047536.1 NR_047537.1 NR_047538.1 NR_047539.1 NR_047540.1 NR_047541.1 NR_047542.1 NR_047543.1 NR_120536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6983462 c.265C>T p.R89W Chr.9: 22006139-22006139 on Build GRCh38
4746788 c.178_236del59 p.R60fs*7 Chr.9: 22006168-22006226 on Build GRCh38
48301 c.178C>T p.R60C Chr.9: 22006226-22006226 on Build GRCh38
4877207 c.207C>T p.G69G Chr.9: 22006197-22006197 on Build GRCh38
4883747 c.250G>T p.V84L Chr.9: 22006154-22006154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764442780 Chr.9: 22006286-22006286 on Build GRCh38
rs777125202 Chr.9: 22006217-22006217 on Build GRCh38
rs927126044 Chr.9: 22006300-22006300 on Build GRCh38
rs765231771 Chr.9: 22006175-22006175 on Build GRCh38
rs979008965 Chr.9: 22006239-22006239 on Build GRCh38
rs551689864 Chr.9: 22006258-22006258 on Build GRCh38
rs746984960 Chr.9: 22006131-22006131 on Build GRCh38
rs1002162923 Chr.9: 22006185-22006185 on Build GRCh38
rs781070051 Chr.9: 22006228-22006228 on Build GRCh38
rs1003403329 Chr.9: 22006250-22006250 on Build GRCh38
rs368412874 Chr.9: 22006190-22006190 on Build GRCh38
rs766769221 Chr.9: 22006165-22006165 on Build GRCh38
rs752926081 Chr.9: 22006176-22006176 on Build GRCh38
rs531754702 Chr.9: 22006230-22006230 on Build GRCh38
rs768407545 Chr.9: 22006252-22006252 on Build GRCh38
rs771322888 Chr.9: 22006262-22006263 on Build GRCh38
rs930595711 Chr.9: 22006307-22006307 on Build GRCh38
rs771295942 Chr.9: 22006259-22006259 on Build GRCh38
rs866457591 Chr.9: 22006138-22006138 on Build GRCh38
rs759975176 Chr.9: 22006218-22006218 on Build GRCh38
rs773400600 Chr.9: 22006157-22006157 on Build GRCh38
rs770734081 Chr.9: 22006139-22006139 on Build GRCh38
rs771488156 Chr.9: 22006215-22006215 on Build GRCh38
rs751280469 Chr.9: 22006182-22006182 on Build GRCh38
rs779655098 Chr.9: 22006199-22006199 on Build GRCh38
rs778420034 Chr.9: 22006256-22006256 on Build GRCh38
rs756996826 Chr.9: 22006227-22006227 on Build GRCh38
rs773670765 Chr.9: 22006211-22006211 on Build GRCh38
rs1007263689 Chr.9: 22006315-22006315 on Build GRCh38
rs988990933 Chr.9: 22006280-22006280 on Build GRCh38
rs745348156 Chr.9: 22006144-22006144 on Build GRCh38
rs776320098 Chr.9: 22006143-22006143 on Build GRCh38
rs752675430 Chr.9: 22006223-22006223 on Build GRCh38
rs762447471 Chr.9: 22006153-22006153 on Build GRCh38
rs768154528 Chr.9: 22006210-22006210 on Build GRCh38
rs142570894 Chr.9: 22006155-22006155 on Build GRCh38
rs779743053 Chr.9: 22006243-22006243 on Build GRCh38
rs769434878 Chr.9: 22006197-22006197 on Build GRCh38
rs144131923 Chr.9: 22006208-22006208 on Build GRCh38
rs761279768 Chr.9: 22006164-22006164 on Build GRCh38
rs775688678 Chr.9: 22006282-22006282 on Build GRCh38
rs370236504 Chr.9: 22006196-22006196 on Build GRCh38
rs368143223 Chr.9: 22006284-22006284 on Build GRCh38
rs748993568 Chr.9: 22006200-22006200 on Build GRCh38
rs776929094 Chr.9: 22006260-22006260 on Build GRCh38
rs764199965 Chr.9: 22006224-22006224 on Build GRCh38
rs759601088 Chr.9: 22006174-22006174 on Build GRCh38
rs779064195 Chr.9: 22006304-22006304 on Build GRCh38
rs967786014 Chr.9: 22006311-22006311 on Build GRCh38
rs752985220 Chr.9: 22006202-22006202 on Build GRCh38
rs909882138 Chr.9: 22006212-22006212 on Build GRCh38
rs949896511 Chr.9: 22006147-22006147 on Build GRCh38
rs565688322 Chr.9: 22006149-22006149 on Build GRCh38
rs2069426 Chr.9: 22006274-22006274 on Build GRCh38
rs3217981 Chr.9: 22006270-22006270 on Build GRCh38
rs751427195 Chr.9: 22006226-22006226 on Build GRCh38
rs148421170 Chr.9: 22006148-22006148 on Build GRCh38
rs536369645 Chr.9: 22006312-22006312 on Build GRCh38
rs757266462 Chr.9: 22006186-22006186 on Build GRCh38
rs753970895 Chr.9: 22006168-22006168 on Build GRCh38
rs760928344 Chr.9: 22006213-22006213 on Build GRCh38
rs1048952968 Chr.9: 22006308-22006308 on Build GRCh38
rs777650997 Chr.9: 22006178-22006178 on Build GRCh38
rs567609105 Chr.9: 22006302-22006302 on Build GRCh38
rs758497193 Chr.9: 22006177-22006177 on Build GRCh38
rs748720669 Chr.9: 22006247-22006247 on Build GRCh38
rs201282617 Chr.9: 22006203-22006203 on Build GRCh38
rs756067582 Chr.9: 22006241-22006241 on Build GRCh38

View Full Product Details