Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84414661-84414900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE2B
Entrez Gene ID
58511
Gene Name
deoxyribonuclease II beta
Gene Aliases
DLAD
UniGene
Hs.129142
Gene Chromosome Location
Chr.1: 84398532-84415895 on Build GRCh38
Transcripts
NM_021233.2 NM_058248.1 XM_011541878.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344546 c.296A>G p.K99R Chr.1: 84414702-84414702 on Build GRCh38
5679357 c.952C>A p.R318S Chr.1: 84414734-84414734 on Build GRCh38
2243639 c.980G>A p.R327Q Chr.1: 84414762-84414762 on Build GRCh38
1661522 c.443A>T p.Y148F Chr.1: 84414849-84414849 on Build GRCh38
3492947 c.924G>A p.W308* Chr.1: 84414706-84414706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760446433 Chr.1: 84414719-84414719 on Build GRCh38
rs751888980 Chr.1: 84414746-84414746 on Build GRCh38
rs889576489 Chr.1: 84414722-84414722 on Build GRCh38
rs994127903 Chr.1: 84414808-84414808 on Build GRCh38
rs771852488 Chr.1: 84414842-84414843 on Build GRCh38
rs145614783 Chr.1: 84414762-84414762 on Build GRCh38
rs777143916 Chr.1: 84414792-84414792 on Build GRCh38
rs771876558 Chr.1: 84414810-84414810 on Build GRCh38
rs182527874 Chr.1: 84414832-84414832 on Build GRCh38
rs773211478 Chr.1: 84414811-84414811 on Build GRCh38
rs557864256 Chr.1: 84414848-84414848 on Build GRCh38
rs199602718 Chr.1: 84414869-84414869 on Build GRCh38
rs776351674 Chr.1: 84414829-84414829 on Build GRCh38
rs763553743 Chr.1: 84414843-84414843 on Build GRCh38
rs753453449 Chr.1: 84414723-84414723 on Build GRCh38
rs570370636 Chr.1: 84414740-84414740 on Build GRCh38
rs764754558 Chr.1: 84414833-84414833 on Build GRCh38
rs202202221 Chr.1: 84414767-84414767 on Build GRCh38
rs767758185 Chr.1: 84414867-84414867 on Build GRCh38
rs781335051 Chr.1: 84414761-84414761 on Build GRCh38
rs147325752 Chr.1: 84414828-84414828 on Build GRCh38
rs146223730 Chr.1: 84414735-84414735 on Build GRCh38
rs753207337 Chr.1: 84414739-84414739 on Build GRCh38
rs766122330 Chr.1: 84414721-84414721 on Build GRCh38
rs746783773 Chr.1: 84414815-84414815 on Build GRCh38
rs750598705 Chr.1: 84414873-84414873 on Build GRCh38
rs570910368 Chr.1: 84414863-84414863 on Build GRCh38
rs751936110 Chr.1: 84414861-84414861 on Build GRCh38
rs3768250 Chr.1: 84414697-84414697 on Build GRCh38
rs933675481 Chr.1: 84414699-84414699 on Build GRCh38
rs76042396 Chr.1: 84414744-84414744 on Build GRCh38
rs191605025 Chr.1: 84414698-84414698 on Build GRCh38
rs373884042 Chr.1: 84414747-84414747 on Build GRCh38
rs770718741 Chr.1: 84414819-84414819 on Build GRCh38
rs565736451 Chr.1: 84414805-84414805 on Build GRCh38
rs771855021 Chr.1: 84414716-84414716 on Build GRCh38
rs759054755 Chr.1: 84414727-84414727 on Build GRCh38
rs1026569429 Chr.1: 84414827-84414827 on Build GRCh38
rs900648170 Chr.1: 84414752-84414752 on Build GRCh38
rs778741286 Chr.1: 84414778-84414778 on Build GRCh38
rs761548011 Chr.1: 84414709-84414709 on Build GRCh38
rs774840549 Chr.1: 84414838-84414838 on Build GRCh38

View Full Product Details