Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169882819-169883048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
UniGene
Hs.586174
Gene Chromosome Location
Chr.5: 169637247-170083382 on Build GRCh38
Transcripts
NM_004946.2 XM_011534449.2 XM_011534451.2 XM_017009190.1 NM_001129891.1 XM_011534415.2 XM_017008908.1

Gene Symbol
FAM196B
Entrez Gene ID
100131897
Gene Name
family with sequence similarity 196 member B
Gene Aliases
C5orf57
UniGene
Hs.721917
Gene Chromosome Location
Chr.5: 169861304-169980740 on Build GRCh38
Transcripts
NM_004946.2 XM_011534449.2 XM_011534451.2 XM_017009190.1 NM_001129891.1 XM_011534415.2 XM_017008908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440517 c.1071G>T p.Q357H Chr.5: 169882828-169882828 on Build GRCh38
4930145 c.1004A>G p.N335S Chr.5: 169882895-169882895 on Build GRCh38
6296292 c.877C>A p.L293M Chr.5: 169883022-169883022 on Build GRCh38
6401217 c.878T>A p.L293Q Chr.5: 169883021-169883021 on Build GRCh38
7192039 c.871G>A p.G291S Chr.5: 169883028-169883028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762880504 Chr.5: 169882948-169882948 on Build GRCh38
rs573686009 Chr.5: 169882932-169882932 on Build GRCh38
rs368906539 Chr.5: 169882999-169882999 on Build GRCh38
rs746974327 Chr.5: 169882857-169882857 on Build GRCh38
rs775947429 Chr.5: 169883007-169883007 on Build GRCh38
rs1038364758 Chr.5: 169883018-169883018 on Build GRCh38
rs201919063 Chr.5: 169882996-169882996 on Build GRCh38
rs956538677 Chr.5: 169882856-169882856 on Build GRCh38
rs540074546 Chr.5: 169882863-169882863 on Build GRCh38
rs1001204172 Chr.5: 169882848-169882848 on Build GRCh38
rs373092434 Chr.5: 169882855-169882855 on Build GRCh38
rs375307639 Chr.5: 169882985-169882985 on Build GRCh38
rs542645445 Chr.5: 169882979-169882979 on Build GRCh38
rs987898741 Chr.5: 169882916-169882916 on Build GRCh38
rs989788665 Chr.5: 169882859-169882859 on Build GRCh38
rs928445393 Chr.5: 169882970-169882970 on Build GRCh38
rs970824776 Chr.5: 169882980-169882980 on Build GRCh38
rs981248621 Chr.5: 169882964-169882964 on Build GRCh38
rs912950568 Chr.5: 169882942-169882942 on Build GRCh38
rs79605350 Chr.5: 169882986-169882986 on Build GRCh38
rs967535325 Chr.5: 169882952-169882952 on Build GRCh38
rs963733929 Chr.5: 169882845-169882845 on Build GRCh38
rs768442702 Chr.5: 169882917-169882917 on Build GRCh38

View Full Product Details