Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27138211-27138436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4831104 c.62C>A p.T21N Chr.2: 27138268-27138268 on Build GRCh38
1690313 c.119C>T p.P40L Chr.2: 27138211-27138211 on Build GRCh38
4532608 c.18G>A p.K6K Chr.2: 27138312-27138312 on Build GRCh38
5693594 c.50C>A p.P17H Chr.2: 27138280-27138280 on Build GRCh38
5695122 c.94C>T p.P32S Chr.2: 27138236-27138236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143780871 Chr.2: 27138331-27138331 on Build GRCh38
rs766337782 Chr.2: 27138248-27138248 on Build GRCh38
rs915588033 Chr.2: 27138384-27138385 on Build GRCh38
rs367907710 Chr.2: 27138296-27138296 on Build GRCh38
rs374970677 Chr.2: 27138280-27138280 on Build GRCh38
rs765974242 Chr.2: 27138346-27138346 on Build GRCh38
rs758466896 Chr.2: 27138246-27138246 on Build GRCh38
rs760470470 Chr.2: 27138330-27138330 on Build GRCh38
rs2304679 Chr.2: 27138354-27138354 on Build GRCh38
rs969345653 Chr.2: 27138295-27138295 on Build GRCh38
rs543742544 Chr.2: 27138305-27138305 on Build GRCh38
rs751688433 Chr.2: 27138353-27138353 on Build GRCh38
rs753140818 Chr.2: 27138365-27138365 on Build GRCh38
rs145535878 Chr.2: 27138277-27138277 on Build GRCh38
rs779428124 Chr.2: 27138394-27138394 on Build GRCh38
rs770519932 Chr.2: 27138302-27138302 on Build GRCh38
rs771927604 Chr.2: 27138328-27138328 on Build GRCh38
rs755532316 Chr.2: 27138272-27138272 on Build GRCh38
rs34036706 Chr.2: 27138319-27138320 on Build GRCh38
rs750492071 Chr.2: 27138245-27138245 on Build GRCh38
rs956964990 Chr.2: 27138260-27138260 on Build GRCh38
rs775275641 Chr.2: 27138329-27138329 on Build GRCh38
rs200670608 Chr.2: 27138326-27138326 on Build GRCh38
rs756539150 Chr.2: 27138366-27138366 on Build GRCh38
rs752161876 Chr.2: 27138261-27138261 on Build GRCh38
rs779346142 Chr.2: 27138316-27138316 on Build GRCh38
rs777438697 Chr.2: 27138282-27138282 on Build GRCh38
rs940414402 Chr.2: 27138416-27138416 on Build GRCh38
rs2304680 Chr.2: 27138378-27138378 on Build GRCh38
rs750923648 Chr.2: 27138352-27138352 on Build GRCh38
rs781734497 Chr.2: 27138275-27138275 on Build GRCh38
rs201696070 Chr.2: 27138358-27138358 on Build GRCh38
rs376148989 Chr.2: 27138395-27138395 on Build GRCh38
rs549359564 Chr.2: 27138383-27138383 on Build GRCh38

View Full Product Details