Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40997331-40997588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001301237.1 NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394659 c.956A>G p.K319R Chr.1: 40997477-40997477 on Build GRCh38
909510 c.873-1G>T p.? Chr.1: 40997393-40997393 on Build GRCh38
1584559 c.924G>A p.T308T Chr.1: 40997445-40997445 on Build GRCh38
6394658 c.956A>G p.K319R Chr.1: 40997477-40997477 on Build GRCh38
5481161 c.931T>A p.S311T Chr.1: 40997452-40997452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758249860 Chr.1: 40997365-40997365 on Build GRCh38
rs755690012 Chr.1: 40997562-40997562 on Build GRCh38
rs568966876 Chr.1: 40997503-40997503 on Build GRCh38
rs765228477 Chr.1: 40997527-40997528 on Build GRCh38
rs201952022 Chr.1: 40997445-40997445 on Build GRCh38
rs780616076 Chr.1: 40997391-40997391 on Build GRCh38
rs560937198 Chr.1: 40997397-40997397 on Build GRCh38
rs765342446 Chr.1: 40997481-40997481 on Build GRCh38
rs200087914 Chr.1: 40997466-40997466 on Build GRCh38
rs946377579 Chr.1: 40997414-40997414 on Build GRCh38
rs750711063 Chr.1: 40997493-40997493 on Build GRCh38
rs754482596 Chr.1: 40997381-40997381 on Build GRCh38
rs372743741 Chr.1: 40997401-40997401 on Build GRCh38
rs776135557 Chr.1: 40997421-40997421 on Build GRCh38
rs754854616 Chr.1: 40997544-40997544 on Build GRCh38
rs772950670 Chr.1: 40997402-40997402 on Build GRCh38
rs756071517 Chr.1: 40997535-40997535 on Build GRCh38
rs201674363 Chr.1: 40997420-40997420 on Build GRCh38
rs781131089 Chr.1: 40997548-40997548 on Build GRCh38
rs902429844 Chr.1: 40997422-40997422 on Build GRCh38
rs761980609 Chr.1: 40997470-40997470 on Build GRCh38
rs1003752907 Chr.1: 40997460-40997460 on Build GRCh38
rs746188198 Chr.1: 40997410-40997410 on Build GRCh38
rs751363108 Chr.1: 40997519-40997519 on Build GRCh38
rs878916397 Chr.1: 40997507-40997507 on Build GRCh38
rs750105098 Chr.1: 40997380-40997380 on Build GRCh38
rs897158288 Chr.1: 40997515-40997515 on Build GRCh38
rs758118584 Chr.1: 40997497-40997497 on Build GRCh38
rs752109334 Chr.1: 40997560-40997560 on Build GRCh38
rs35240983 Chr.1: 40997479-40997479 on Build GRCh38
rs1020095716 Chr.1: 40997457-40997457 on Build GRCh38
rs779949097 Chr.1: 40997372-40997372 on Build GRCh38

View Full Product Details