Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28308762-28309035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT1
Entrez Gene ID
2321
Gene Name
fms related tyrosine kinase 1
Gene Aliases
FLT, FLT-1, VEGFR-1, VEGFR1
UniGene
Hs.594454
Gene Chromosome Location
Chr.13: 28300346-28495128 on Build GRCh38
Transcripts
NM_002019.4 XM_017020485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928345 c.3636-1G>C p.? Chr.13: 28308928-28308928 on Build GRCh38
3885168 c.3671G>A p.R1224K Chr.13: 28308892-28308892 on Build GRCh38
131727 c.3667G>A p.E1223K Chr.13: 28308896-28308896 on Build GRCh38
4784282 c.724T>A p.F242I Chr.13: 28308908-28308908 on Build GRCh38
5376594 c.3699G>A p.P1233P Chr.13: 28308864-28308864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765007272 Chr.13: 28308798-28308799 on Build GRCh38
rs779359571 Chr.13: 28308900-28308900 on Build GRCh38
rs749167073 Chr.13: 28308948-28308948 on Build GRCh38
rs775361050 Chr.13: 28308818-28308818 on Build GRCh38
rs758710745 Chr.13: 28308923-28308923 on Build GRCh38
rs574300641 Chr.13: 28308821-28308821 on Build GRCh38
rs762770891 Chr.13: 28308820-28308820 on Build GRCh38
rs770960653 Chr.13: 28308935-28308935 on Build GRCh38
rs761584349 Chr.13: 28308836-28308836 on Build GRCh38
rs369898762 Chr.13: 28308944-28308944 on Build GRCh38
rs767248226 Chr.13: 28308967-28308967 on Build GRCh38
rs76556213 Chr.13: 28308955-28308955 on Build GRCh38
rs773395159 Chr.13: 28308809-28308809 on Build GRCh38
rs115595062 Chr.13: 28308865-28308865 on Build GRCh38
rs967383568 Chr.13: 28308909-28308909 on Build GRCh38
rs867799392 Chr.13: 28308859-28308859 on Build GRCh38
rs769776997 Chr.13: 28308814-28308814 on Build GRCh38
rs9579177 Chr.13: 28308811-28308811 on Build GRCh38
rs373145963 Chr.13: 28308941-28308941 on Build GRCh38
rs561054355 Chr.13: 28308932-28308932 on Build GRCh38
rs990680025 Chr.13: 28308988-28308988 on Build GRCh38
rs748333380 Chr.13: 28308799-28308799 on Build GRCh38
rs61763567 Chr.13: 28309003-28309003 on Build GRCh38
rs61763568 Chr.13: 28308990-28308990 on Build GRCh38
rs577092483 Chr.13: 28308788-28308788 on Build GRCh38
rs779306159 Chr.13: 28308793-28308793 on Build GRCh38
rs376553481 Chr.13: 28308864-28308864 on Build GRCh38
rs758313026 Chr.13: 28308912-28308914 on Build GRCh38
rs545962874 Chr.13: 28308806-28308806 on Build GRCh38
rs377496044 Chr.13: 28308959-28308959 on Build GRCh38
rs755396275 Chr.13: 28308897-28308897 on Build GRCh38
rs370986273 Chr.13: 28308892-28308892 on Build GRCh38
rs369151368 Chr.13: 28308808-28308808 on Build GRCh38
rs768570374 Chr.13: 28308950-28308950 on Build GRCh38
rs1035738787 Chr.13: 28308927-28308927 on Build GRCh38
rs767014384 Chr.13: 28308871-28308871 on Build GRCh38
rs767072926 Chr.13: 28308851-28308851 on Build GRCh38
rs750340870 Chr.13: 28308842-28308842 on Build GRCh38
rs975374800 Chr.13: 28309010-28309010 on Build GRCh38
rs7993418 Chr.13: 28308924-28308924 on Build GRCh38
rs761480667 Chr.13: 28308961-28308961 on Build GRCh38
rs773891087 Chr.13: 28308835-28308835 on Build GRCh38

View Full Product Details