Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 2819399-2819658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN4
Entrez Gene ID
152330
Gene Name
contactin 4
Gene Aliases
AXCAM, BIG-2
UniGene
Hs.298705
Gene Chromosome Location
Chr.3: 2098771-3059080 on Build GRCh38
Transcripts
NM_001206955.1 NM_175607.2 XM_006713004.3 XM_011533425.2 XM_011533426.2 XM_011533427.2 XM_011533428.2 XM_011533429.2 XM_011533430.2 XM_011533431.2 XM_017005782.1 XM_017005783.1 XM_017005784.1 XM_017005785.1 XM_017005786.1 XM_017005787.1 XM_017005788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1422287 c.437C>T p.P146L Chr.3: 2819564-2819564 on Build GRCh38
3592258 c.416G>A p.G139E Chr.3: 2819543-2819543 on Build GRCh38
2983350 c.407G>A p.R136Q Chr.3: 2819534-2819534 on Build GRCh38
3392328 c.404G>A p.R135H Chr.3: 2819531-2819531 on Build GRCh38
730369 c.380G>T p.R127I Chr.3: 2819507-2819507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576375620 Chr.3: 2819534-2819534 on Build GRCh38
rs1012598240 Chr.3: 2819487-2819487 on Build GRCh38
rs878949592 Chr.3: 2819491-2819491 on Build GRCh38
rs752325056 Chr.3: 2819586-2819586 on Build GRCh38
rs753579463 Chr.3: 2819616-2819616 on Build GRCh38
rs749173059 Chr.3: 2819486-2819486 on Build GRCh38
rs987280347 Chr.3: 2819509-2819509 on Build GRCh38
rs187134406 Chr.3: 2819626-2819626 on Build GRCh38
rs372398969 Chr.3: 2819620-2819620 on Build GRCh38
rs1022600859 Chr.3: 2819502-2819502 on Build GRCh38
rs999945370 Chr.3: 2819477-2819477 on Build GRCh38
rs1035465281 Chr.3: 2819458-2819458 on Build GRCh38
rs767055199 Chr.3: 2819585-2819585 on Build GRCh38
rs147652210 Chr.3: 2819480-2819480 on Build GRCh38
rs752415074 Chr.3: 2819470-2819470 on Build GRCh38
rs762319356 Chr.3: 2819553-2819553 on Build GRCh38
rs371852870 Chr.3: 2819522-2819522 on Build GRCh38
rs751039356 Chr.3: 2819443-2819443 on Build GRCh38
rs868306780 Chr.3: 2819533-2819533 on Build GRCh38
rs754602812 Chr.3: 2819452-2819452 on Build GRCh38
rs755783590 Chr.3: 2819594-2819594 on Build GRCh38
rs368010840 Chr.3: 2819454-2819454 on Build GRCh38
rs757172793 Chr.3: 2819490-2819490 on Build GRCh38
rs374294005 Chr.3: 2819438-2819438 on Build GRCh38
rs758964447 Chr.3: 2819574-2819574 on Build GRCh38
rs772247450 Chr.3: 2819523-2819523 on Build GRCh38
rs374705571 Chr.3: 2819436-2819436 on Build GRCh38
rs141541671 Chr.3: 2819550-2819550 on Build GRCh38
rs778698307 Chr.3: 2819621-2819621 on Build GRCh38
rs377113483 Chr.3: 2819531-2819531 on Build GRCh38
rs765679434 Chr.3: 2819554-2819554 on Build GRCh38
rs746320849 Chr.3: 2819624-2819624 on Build GRCh38
rs368759954 Chr.3: 2819437-2819437 on Build GRCh38
rs977295576 Chr.3: 2819564-2819564 on Build GRCh38
rs773828221 Chr.3: 2819569-2819569 on Build GRCh38
rs777592806 Chr.3: 2819484-2819484 on Build GRCh38
rs201133857 Chr.3: 2819496-2819496 on Build GRCh38
rs763834252 Chr.3: 2819612-2819612 on Build GRCh38
rs775849107 Chr.3: 2819530-2819530 on Build GRCh38

View Full Product Details