Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6222285-6222528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362855 c.1155C>T p.L385L Chr.11: 6222478-6222478 on Build GRCh38
5665663 c.1128G>A p.L376L Chr.11: 6222505-6222505 on Build GRCh38
4682650 c.1189C>T p.R397W Chr.11: 6222444-6222444 on Build GRCh38
5955599 c.1128G>T p.L376F Chr.11: 6222505-6222505 on Build GRCh38
6387615 c.1119C>A p.F373L Chr.11: 6222514-6222514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746620304 Chr.11: 6222461-6222461 on Build GRCh38
rs372557789 Chr.11: 6222473-6222473 on Build GRCh38
rs916316009 Chr.11: 6222405-6222405 on Build GRCh38
rs771687836 Chr.11: 6222456-6222456 on Build GRCh38
rs200972055 Chr.11: 6222469-6222469 on Build GRCh38
rs755174314 Chr.11: 6222489-6222489 on Build GRCh38
rs170524 Chr.11: 6222408-6222408 on Build GRCh38
rs763912758 Chr.11: 6222432-6222433 on Build GRCh38
rs778417119 Chr.11: 6222433-6222433 on Build GRCh38
rs1019158633 Chr.11: 6222319-6222319 on Build GRCh38
rs371662276 Chr.11: 6222444-6222444 on Build GRCh38
rs966329869 Chr.11: 6222370-6222370 on Build GRCh38
rs535861381 Chr.11: 6222421-6222421 on Build GRCh38
rs371497441 Chr.11: 6222460-6222460 on Build GRCh38
rs918852443 Chr.11: 6222496-6222496 on Build GRCh38
rs761507239 Chr.11: 6222466-6222466 on Build GRCh38
rs557347285 Chr.11: 6222459-6222459 on Build GRCh38
rs770614230 Chr.11: 6222419-6222419 on Build GRCh38
rs756573925 Chr.11: 6222430-6222430 on Build GRCh38
rs993775413 Chr.11: 6222353-6222353 on Build GRCh38
rs753176538 Chr.11: 6222500-6222500 on Build GRCh38
rs983865628 Chr.11: 6222380-6222380 on Build GRCh38
rs113400294 Chr.11: 6222329-6222329 on Build GRCh38
rs776419744 Chr.11: 6222465-6222465 on Build GRCh38
rs766402546 Chr.11: 6222478-6222478 on Build GRCh38
rs751798951 Chr.11: 6222483-6222483 on Build GRCh38
rs765275651 Chr.11: 6222468-6222468 on Build GRCh38
rs200051589 Chr.11: 6222377-6222377 on Build GRCh38
rs145458032 Chr.11: 6222462-6222462 on Build GRCh38
rs756424921 Chr.11: 6222501-6222501 on Build GRCh38
rs781575569 Chr.11: 6222417-6222417 on Build GRCh38
rs745431912 Chr.11: 6222443-6222443 on Build GRCh38
rs763528961 Chr.11: 6222426-6222426 on Build GRCh38
rs767781991 Chr.11: 6222499-6222499 on Build GRCh38
rs1020889900 Chr.11: 6222333-6222333 on Build GRCh38
rs753136466 Chr.11: 6222425-6222425 on Build GRCh38

View Full Product Details