Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43793590-43793863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR1282
Entrez Gene ID
100302254
Gene Name
microRNA 1282
Gene Aliases
MIRN1282, hsa-mir-1282
UniGene
Gene Chromosome Location
Chr.15: 43793659-43793759 on Build GRCh38
Transcripts
NR_031695.1 NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1

Gene Symbol
SERF2
Entrez Gene ID
10169
Gene Name
small EDRK-rich factor 2
Gene Aliases
4F5REL, FAM2C, H4F5REL, HsT17089
UniGene
Hs.424126
Gene Chromosome Location
Chr.15: 43777096-43796089 on Build GRCh38
Transcripts
NR_031695.1 NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1

Gene Symbol
SERF2-C15ORF63
Entrez Gene ID
100529067
Gene Name
SERF2-C15orf63 readthrough
Gene Aliases
-
UniGene
Hs.730672
Gene Chromosome Location
Chr.15: 43791976-43802589 on Build GRCh38
Transcripts
NR_031695.1 NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433934 c.117-10C>G p.? Chr.15: 43793700-43793700 on Build GRCh38
6322038 c.229G>T p.A77S Chr.15: 43793688-43793688 on Build GRCh38
1478147 c.241C>G p.L81V Chr.15: 43793700-43793700 on Build GRCh38
3981516 c.256C>A p.R86R Chr.15: 43793715-43793715 on Build GRCh38
3981517 c.122C>A p.S41* Chr.15: 43793715-43793715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749008549 Chr.15: 43793826-43793826 on Build GRCh38
rs538690173 Chr.15: 43793656-43793656 on Build GRCh38
rs776224437 Chr.15: 43793786-43793787 on Build GRCh38
rs750512118 Chr.15: 43793818-43793818 on Build GRCh38
rs757357108 Chr.15: 43793680-43793680 on Build GRCh38
rs764238650 Chr.15: 43793809-43793809 on Build GRCh38
rs918571408 Chr.15: 43793621-43793621 on Build GRCh38
rs775913330 Chr.15: 43793628-43793629 on Build GRCh38
rs11555370 Chr.15: 43793834-43793834 on Build GRCh38
rs745342456 Chr.15: 43793835-43793835 on Build GRCh38
rs369832525 Chr.15: 43793684-43793684 on Build GRCh38
rs764915359 Chr.15: 43793828-43793829 on Build GRCh38
rs770557996 Chr.15: 43793830-43793830 on Build GRCh38
rs558401790 Chr.15: 43793670-43793670 on Build GRCh38
rs367904215 Chr.15: 43793661-43793661 on Build GRCh38
rs112115773 Chr.15: 43793820-43793820 on Build GRCh38
rs768037613 Chr.15: 43793803-43793803 on Build GRCh38
rs879249287 Chr.15: 43793686-43793686 on Build GRCh38
rs761280384 Chr.15: 43793814-43793815 on Build GRCh38
rs11269 Chr.15: 43793711-43793711 on Build GRCh38
rs553559156 Chr.15: 43793619-43793619 on Build GRCh38
rs575152814 Chr.15: 43793811-43793811 on Build GRCh38
rs1045969535 Chr.15: 43793640-43793640 on Build GRCh38
rs11555372 Chr.15: 43793817-43793817 on Build GRCh38
rs772478588 Chr.15: 43793836-43793836 on Build GRCh38
rs369902877 Chr.15: 43793813-43793813 on Build GRCh38
rs573381677 Chr.15: 43793628-43793628 on Build GRCh38
rs760950472 Chr.15: 43793841-43793841 on Build GRCh38
rs769462084 Chr.15: 43793618-43793618 on Build GRCh38
rs754908367 Chr.15: 43793821-43793821 on Build GRCh38
rs747914951 Chr.15: 43793823-43793823 on Build GRCh38
rs761170966 Chr.15: 43793702-43793702 on Build GRCh38
rs772869968 Chr.15: 43793783-43793783 on Build GRCh38
rs763552438 Chr.15: 43793751-43793753 on Build GRCh38
rs747940819 Chr.15: 43793767-43793767 on Build GRCh38
rs201274196 Chr.15: 43793786-43793786 on Build GRCh38
rs769358781 Chr.15: 43793778-43793778 on Build GRCh38
rs375164328 Chr.15: 43793647-43793647 on Build GRCh38
rs766891421 Chr.15: 43793631-43793631 on Build GRCh38
rs752003521 Chr.15: 43793638-43793638 on Build GRCh38
rs770359655 Chr.15: 43793787-43793787 on Build GRCh38
rs1802422 Chr.15: 43793838-43793838 on Build GRCh38
rs776005780 Chr.15: 43793840-43793840 on Build GRCh38
rs753035270 Chr.15: 43793804-43793804 on Build GRCh38
rs772634038 Chr.15: 43793625-43793625 on Build GRCh38
rs1043887061 Chr.15: 43793819-43793819 on Build GRCh38
rs753092831 Chr.15: 43793653-43793653 on Build GRCh38
rs755878719 Chr.15: 43793824-43793824 on Build GRCh38
rs773900330 Chr.15: 43793833-43793833 on Build GRCh38
rs962753506 Chr.15: 43793611-43793611 on Build GRCh38
rs759897421 Chr.15: 43793639-43793639 on Build GRCh38
rs372561860 Chr.15: 43793689-43793689 on Build GRCh38
rs762406090 Chr.15: 43793629-43793629 on Build GRCh38
rs760093532 Chr.15: 43793794-43793794 on Build GRCh38
rs773845378 Chr.15: 43793793-43793793 on Build GRCh38
rs780903077 Chr.15: 43793695-43793695 on Build GRCh38
rs768705094 Chr.15: 43793690-43793690 on Build GRCh38
rs753510458 Chr.15: 43793807-43793807 on Build GRCh38
rs765474877 Chr.15: 43793815-43793815 on Build GRCh38
rs772482463 Chr.15: 43793619-43793620 on Build GRCh38
rs756331229 Chr.15: 43793658-43793658 on Build GRCh38

View Full Product Details