Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2184400-2184659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
UniGene
Hs.107101
Gene Chromosome Location
Chr.1: 2184460-2212720 on Build GRCh38
Transcripts
NM_001146310.1 NM_001256945.1 NM_001282671.1 NM_001282672.1 NM_001282673.1 XM_017000554.1 NM_001033581.1 NM_001033582.1 NM_001242874.1 NM_002744.4 XM_017001790.1 XM_017001791.1 XM_017001796.1 XM_017001798.1 XM_017001799.1 XM_017001801.1 XM_017001804.1 XM_017001805.1

Gene Symbol
PRKCZ
Entrez Gene ID
5590
Gene Name
protein kinase C zeta
Gene Aliases
PKC-ZETA, PKC2
UniGene
Hs.496255
Gene Chromosome Location
Chr.1: 2050470-2185395 on Build GRCh38
Transcripts
NM_001146310.1 NM_001256945.1 NM_001282671.1 NM_001282672.1 NM_001282673.1 XM_017000554.1 NM_001033581.1 NM_001033582.1 NM_001242874.1 NM_002744.4 XM_017001790.1 XM_017001791.1 XM_017001796.1 XM_017001798.1 XM_017001799.1 XM_017001801.1 XM_017001804.1 XM_017001805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678733 c.1633C>A p.L545M Chr.1: 2184640-2184640 on Build GRCh38
7278498 c.1606C>T p.Q536* Chr.1: 2184613-2184613 on Build GRCh38
4629881 c.1630G>A p.G544S Chr.1: 2184637-2184637 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs143763676 Chr.1: 2184631-2184631 on Build GRCh38
rs568098207 Chr.1: 2184478-2184478 on Build GRCh38
rs997633526 Chr.1: 2184532-2184532 on Build GRCh38
rs370078851 Chr.1: 2184515-2184515 on Build GRCh38
rs990885826 Chr.1: 2184463-2184463 on Build GRCh38
rs760358797 Chr.1: 2184547-2184547 on Build GRCh38
rs1047449400 Chr.1: 2184513-2184513 on Build GRCh38
rs780482466 Chr.1: 2184572-2184572 on Build GRCh38
rs749692865 Chr.1: 2184585-2184585 on Build GRCh38
rs918956967 Chr.1: 2184592-2184592 on Build GRCh38
rs948530990 Chr.1: 2184477-2184477 on Build GRCh38
rs751681990 Chr.1: 2184557-2184557 on Build GRCh38
rs772703489 Chr.1: 2184615-2184615 on Build GRCh38
rs553946128 Chr.1: 2184517-2184517 on Build GRCh38
rs554637318 Chr.1: 2184636-2184636 on Build GRCh38
rs930279604 Chr.1: 2184594-2184594 on Build GRCh38
rs766230396 Chr.1: 2184548-2184548 on Build GRCh38
rs781542404 Chr.1: 2184562-2184562 on Build GRCh38
rs539390535 Chr.1: 2184564-2184566 on Build GRCh38
rs756392599 Chr.1: 2184569-2184569 on Build GRCh38
rs769147141 Chr.1: 2184591-2184591 on Build GRCh38
rs775616525 Chr.1: 2184633-2184633 on Build GRCh38
rs45536443 Chr.1: 2184550-2184550 on Build GRCh38
rs141442853 Chr.1: 2184469-2184469 on Build GRCh38
rs1040228811 Chr.1: 2184437-2184437 on Build GRCh38
rs377447072 Chr.1: 2184630-2184630 on Build GRCh38
rs771614705 Chr.1: 2184603-2184603 on Build GRCh38
rs1032319092 Chr.1: 2184442-2184442 on Build GRCh38
rs1033205873 Chr.1: 2184502-2184502 on Build GRCh38
rs928981675 Chr.1: 2184503-2184503 on Build GRCh38
rs528467456 Chr.1: 2184467-2184468 on Build GRCh38
rs199841488 Chr.1: 2184559-2184559 on Build GRCh38
rs190090688 Chr.1: 2184544-2184544 on Build GRCh38
rs928521964 Chr.1: 2184483-2184483 on Build GRCh38
rs772971154 Chr.1: 2184546-2184546 on Build GRCh38
rs958376916 Chr.1: 2184460-2184460 on Build GRCh38
rs966716295 Chr.1: 2184435-2184435 on Build GRCh38
rs113629589 Chr.1: 2184480-2184480 on Build GRCh38
rs779455788 Chr.1: 2184597-2184597 on Build GRCh38
rs958023599 Chr.1: 2184606-2184606 on Build GRCh38
rs1038742032 Chr.1: 2184523-2184523 on Build GRCh38
rs779840498 Chr.1: 2184525-2184525 on Build GRCh38
rs373358003 Chr.1: 2184549-2184549 on Build GRCh38
rs757469641 Chr.1: 2184560-2184560 on Build GRCh38
rs1020302441 Chr.1: 2184445-2184445 on Build GRCh38
rs377346377 Chr.1: 2184543-2184543 on Build GRCh38
rs764199003 Chr.1: 2184551-2184551 on Build GRCh38
rs760400823 Chr.1: 2184621-2184621 on Build GRCh38
rs765251717 Chr.1: 2184632-2184632 on Build GRCh38
rs192915352 Chr.1: 2184566-2184566 on Build GRCh38
rs373967833 Chr.1: 2184527-2184527 on Build GRCh38
rs116090142 Chr.1: 2184441-2184441 on Build GRCh38
rs761527109 Chr.1: 2184573-2184573 on Build GRCh38
rs768098125 Chr.1: 2184535-2184535 on Build GRCh38
rs749035122 Chr.1: 2184599-2184599 on Build GRCh38

View Full Product Details