Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80649967-80650240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHFR
Entrez Gene ID
1719
Gene Name
dihydrofolate reductase
Gene Aliases
DHFRP1, DYR
UniGene
Hs.592364
Gene Chromosome Location
Chr.5: 80626226-80654981 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1 NM_001190470.1

Gene Symbol
MTRNR2L2
Entrez Gene ID
100462981
Gene Name
MT-RNR2-like 2
Gene Aliases
HN2
UniGene
Hs.666077
Gene Chromosome Location
Chr.5: 80650000-80651035 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1 NM_001190470.1

SNP Detail

SNP ID Chromosome Location
rs1001341936 Chr.5: 80650152-80650152 on Build GRCh38
rs892903197 Chr.5: 80650170-80650170 on Build GRCh38
rs545906985 Chr.5: 80649993-80649993 on Build GRCh38
rs374065833 Chr.5: 80650178-80650178 on Build GRCh38
rs557069457 Chr.5: 80650151-80650151 on Build GRCh38
rs767396956 Chr.5: 80650030-80650030 on Build GRCh38
rs755555577 Chr.5: 80650113-80650113 on Build GRCh38
rs905675775 Chr.5: 80650149-80650149 on Build GRCh38
rs763886697 Chr.5: 80649994-80649994 on Build GRCh38
rs78893240 Chr.5: 80650193-80650193 on Build GRCh38
rs1009895794 Chr.5: 80650171-80650171 on Build GRCh38
rs997736024 Chr.5: 80650191-80650191 on Build GRCh38
rs201935861 Chr.5: 80650176-80650177 on Build GRCh38
rs755655550 Chr.5: 80650061-80650061 on Build GRCh38
rs1020411583 Chr.5: 80650173-80650173 on Build GRCh38
rs1045629993 Chr.5: 80650146-80650146 on Build GRCh38
rs779680477 Chr.5: 80650097-80650097 on Build GRCh38
rs199844766 Chr.5: 80650056-80650056 on Build GRCh38
rs537498428 Chr.5: 80650148-80650148 on Build GRCh38
rs75397552 Chr.5: 80650195-80650195 on Build GRCh38
rs536766880 Chr.5: 80650208-80650208 on Build GRCh38
rs10942928 Chr.5: 80650052-80650052 on Build GRCh38
rs772333041 Chr.5: 80650174-80650174 on Build GRCh38
rs761738663 Chr.5: 80650029-80650029 on Build GRCh38
rs573937678 Chr.5: 80650154-80650154 on Build GRCh38
rs571826098 Chr.5: 80650115-80650115 on Build GRCh38
rs80109921 Chr.5: 80650169-80650169 on Build GRCh38
rs528078048 Chr.5: 80650014-80650014 on Build GRCh38
rs551689879 Chr.5: 80650076-80650076 on Build GRCh38
rs748611781 Chr.5: 80650134-80650134 on Build GRCh38
rs753272540 Chr.5: 80650108-80650108 on Build GRCh38
rs78064912 Chr.5: 80650162-80650162 on Build GRCh38
rs976247774 Chr.5: 80650040-80650040 on Build GRCh38
rs918504266 Chr.5: 80650144-80650144 on Build GRCh38
rs953536177 Chr.5: 80650071-80650071 on Build GRCh38
rs760940508 Chr.5: 80650058-80650061 on Build GRCh38
rs781614421 Chr.5: 80650160-80650161 on Build GRCh38
rs772444192 Chr.5: 80650135-80650135 on Build GRCh38
rs949967255 Chr.5: 80650145-80650145 on Build GRCh38
rs74561694 Chr.5: 80650094-80650094 on Build GRCh38

View Full Product Details