Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108222381-108222654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATM
Entrez Gene ID
472
Gene Name
ATM serine/threonine kinase
Gene Aliases
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
UniGene
Hs.367437
Gene Chromosome Location
Chr.11: 108222484-108369102 on Build GRCh38
Transcripts
XM_011542843.2 XM_017017791.1 XM_017017792.1 NM_001321307.1 NM_002519.2 XM_011542854.2

Gene Symbol
NPAT
Entrez Gene ID
4863
Gene Name
nuclear protein, coactivator of histone transcription
Gene Aliases
E14, E14/NPAT, p220
UniGene
Hs.171061
Gene Chromosome Location
Chr.11: 108155280-108222638 on Build GRCh38
Transcripts
XM_011542843.2 XM_017017791.1 XM_017017792.1 NM_001321307.1 NM_002519.2 XM_011542854.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207211 c.2_3insATCATG p.M1>ISW Chr.11: 108222534-108222535 on Build GRCh38
7380128 c.10C>T p.P4S Chr.11: 108222527-108222527 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987030345 Chr.11: 108222436-108222436 on Build GRCh38
rs758245923 Chr.11: 108222540-108222540 on Build GRCh38
rs1056793846 Chr.11: 108222574-108222574 on Build GRCh38
rs80163666 Chr.11: 108222434-108222434 on Build GRCh38
rs773550815 Chr.11: 108222471-108222471 on Build GRCh38
rs780092648 Chr.11: 108222554-108222554 on Build GRCh38
rs865795034 Chr.11: 108222633-108222633 on Build GRCh38
rs763635146 Chr.11: 108222507-108222507 on Build GRCh38
rs537878674 Chr.11: 108222528-108222528 on Build GRCh38
rs770600720 Chr.11: 108222529-108222531 on Build GRCh38
rs1028476984 Chr.11: 108222423-108222423 on Build GRCh38
rs768802463 Chr.11: 108222566-108222566 on Build GRCh38
rs779009909 Chr.11: 108222530-108222530 on Build GRCh38
rs534566260 Chr.11: 108222616-108222616 on Build GRCh38
rs1009896202 Chr.11: 108222596-108222596 on Build GRCh38
rs942455067 Chr.11: 108222632-108222632 on Build GRCh38
rs228589 Chr.11: 108222481-108222481 on Build GRCh38
rs753469498 Chr.11: 108222512-108222512 on Build GRCh38
rs774623796 Chr.11: 108222579-108222579 on Build GRCh38
rs900898578 Chr.11: 108222634-108222634 on Build GRCh38
rs376542079 Chr.11: 108222585-108222585 on Build GRCh38
rs547339680 Chr.11: 108222628-108222628 on Build GRCh38
rs948814410 Chr.11: 108222504-108222504 on Build GRCh38
rs76719341 Chr.11: 108222586-108222586 on Build GRCh38
rs905047784 Chr.11: 108222527-108222527 on Build GRCh38
rs199579930 Chr.11: 108222573-108222573 on Build GRCh38
rs573131053 Chr.11: 108222432-108222432 on Build GRCh38
rs375988543 Chr.11: 108222433-108222433 on Build GRCh38
rs781244000 Chr.11: 108222567-108222567 on Build GRCh38
rs910883026 Chr.11: 108222629-108222629 on Build GRCh38
rs773712159 Chr.11: 108222571-108222571 on Build GRCh38
rs757251547 Chr.11: 108222526-108222526 on Build GRCh38
rs373553992 Chr.11: 108222454-108222454 on Build GRCh38
rs141298949 Chr.11: 108222451-108222451 on Build GRCh38
rs1028004464 Chr.11: 108222627-108222627 on Build GRCh38
rs919916941 Chr.11: 108222492-108222492 on Build GRCh38
rs988640098 Chr.11: 108222606-108222606 on Build GRCh38
rs764587580 Chr.11: 108222599-108222599 on Build GRCh38
rs370264347 Chr.11: 108222595-108222595 on Build GRCh38
rs376250464 Chr.11: 108222459-108222459 on Build GRCh38
rs747232472 Chr.11: 108222558-108222558 on Build GRCh38
rs986951413 Chr.11: 108222635-108222635 on Build GRCh38
rs892344115 Chr.11: 108222576-108222576 on Build GRCh38
rs184150429 Chr.11: 108222611-108222611 on Build GRCh38
rs960127392 Chr.11: 108222597-108222597 on Build GRCh38
rs770226568 Chr.11: 108222570-108222570 on Build GRCh38
rs763320013 Chr.11: 108222475-108222475 on Build GRCh38
rs530679889 Chr.11: 108222602-108222602 on Build GRCh38
rs750178906 Chr.11: 108222537-108222537 on Build GRCh38
rs191855201 Chr.11: 108222607-108222607 on Build GRCh38
rs752422586 Chr.11: 108222485-108222485 on Build GRCh38
rs760050363 Chr.11: 108222488-108222488 on Build GRCh38
rs770353266 Chr.11: 108222464-108222464 on Build GRCh38
rs748284888 Chr.11: 108222568-108222568 on Build GRCh38
rs1044482753 Chr.11: 108222517-108222517 on Build GRCh38
rs747022060 Chr.11: 108222453-108222453 on Build GRCh38

View Full Product Details