Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108088665-108088938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4A
Entrez Gene ID
115201
Gene Name
autophagy related 4A cysteine peptidase
Gene Aliases
APG4A, AUTL2
UniGene
Hs.8763
Gene Chromosome Location
Chr.X: 108088900-108154671 on Build GRCh38
Transcripts
XM_011530841.2 NM_002814.3 NM_170750.2

Gene Symbol
PSMD10
Entrez Gene ID
5716
Gene Name
proteasome 26S subunit, non-ATPase 10
Gene Aliases
dJ889N15.2, p28, p28(GANK)
UniGene
Hs.522752
Gene Chromosome Location
Chr.X: 108084205-108091644 on Build GRCh38
Transcripts
XM_011530841.2 NM_002814.3 NM_170750.2

SNP Detail

SNP ID Chromosome Location
rs770057550 Chr.X: 108088885-108088885 on Build GRCh38
rs943987828 Chr.X: 108088806-108088806 on Build GRCh38
rs749852206 Chr.X: 108088899-108088899 on Build GRCh38
rs765167396 Chr.X: 108088709-108088709 on Build GRCh38
rs11541112 Chr.X: 108088758-108088758 on Build GRCh38
rs757979326 Chr.X: 108088772-108088772 on Build GRCh38
rs757959120 Chr.X: 108088711-108088711 on Build GRCh38
rs920958672 Chr.X: 108088890-108088890 on Build GRCh38
rs776058850 Chr.X: 108088891-108088891 on Build GRCh38
rs201894843 Chr.X: 108088904-108088904 on Build GRCh38
rs11541113 Chr.X: 108088774-108088774 on Build GRCh38
rs780727553 Chr.X: 108088861-108088861 on Build GRCh38
rs777486130 Chr.X: 108088773-108088773 on Build GRCh38
rs146134320 Chr.X: 108088755-108088755 on Build GRCh38
rs746183165 Chr.X: 108088870-108088870 on Build GRCh38
rs369553197 Chr.X: 108088845-108088845 on Build GRCh38
rs1010019712 Chr.X: 108088694-108088694 on Build GRCh38
rs968407342 Chr.X: 108088886-108088886 on Build GRCh38
rs768699439 Chr.X: 108088902-108088902 on Build GRCh38
rs375100790 Chr.X: 108088792-108088792 on Build GRCh38

View Full Product Details