Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35797114-35797367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPS
Entrez Gene ID
1208
Gene Name
colipase
Gene Aliases
-
UniGene
Hs.1340
Gene Chromosome Location
Chr.6: 35794982-35797344 on Build GRCh38
Transcripts
NM_001252597.1 NM_001252598.1 NM_001832.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6174200 c.33C>A p.A11A Chr.6: 35797256-35797256 on Build GRCh38
3994913 c.65G>C p.R22P Chr.6: 35797224-35797224 on Build GRCh38
3921581 c.61C>T p.P21S Chr.6: 35797228-35797228 on Build GRCh38
6513878 c.13C>T p.L5L Chr.6: 35797276-35797276 on Build GRCh38
172140 c.31G>A p.A11T Chr.6: 35797258-35797258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747107279 Chr.6: 35797325-35797325 on Build GRCh38
rs376600832 Chr.6: 35797167-35797167 on Build GRCh38
rs763736793 Chr.6: 35797230-35797230 on Build GRCh38
rs775893258 Chr.6: 35797294-35797294 on Build GRCh38
rs781156085 Chr.6: 35797181-35797181 on Build GRCh38
rs745617742 Chr.6: 35797182-35797182 on Build GRCh38
rs200244589 Chr.6: 35797246-35797246 on Build GRCh38
rs766826077 Chr.6: 35797204-35797204 on Build GRCh38
rs755917684 Chr.6: 35797259-35797259 on Build GRCh38
rs577828670 Chr.6: 35797165-35797165 on Build GRCh38
rs1040553250 Chr.6: 35797303-35797303 on Build GRCh38
rs762563597 Chr.6: 35797295-35797295 on Build GRCh38
rs372093288 Chr.6: 35797225-35797225 on Build GRCh38
rs771434902 Chr.6: 35797277-35797277 on Build GRCh38
rs112222376 Chr.6: 35797235-35797235 on Build GRCh38
rs777068073 Chr.6: 35797288-35797288 on Build GRCh38
rs748386448 Chr.6: 35797190-35797190 on Build GRCh38
rs3748051 Chr.6: 35797166-35797166 on Build GRCh38
rs757230949 Chr.6: 35797175-35797175 on Build GRCh38
rs747055917 Chr.6: 35797275-35797275 on Build GRCh38
rs751119258 Chr.6: 35797298-35797298 on Build GRCh38
rs566561357 Chr.6: 35797319-35797319 on Build GRCh38
rs761365160 Chr.6: 35797266-35797268 on Build GRCh38
rs768924929 Chr.6: 35797183-35797183 on Build GRCh38
rs761215824 Chr.6: 35797199-35797199 on Build GRCh38
rs3748050 Chr.6: 35797291-35797291 on Build GRCh38
rs1054340429 Chr.6: 35797226-35797226 on Build GRCh38
rs776998766 Chr.6: 35797166-35797166 on Build GRCh38
rs201152078 Chr.6: 35797337-35797337 on Build GRCh38
rs758654606 Chr.6: 35797316-35797316 on Build GRCh38
rs747010477 Chr.6: 35797173-35797173 on Build GRCh38
rs199655623 Chr.6: 35797317-35797317 on Build GRCh38
rs755433404 Chr.6: 35797198-35797198 on Build GRCh38
rs78476830 Chr.6: 35797329-35797329 on Build GRCh38
rs2766597 Chr.6: 35797266-35797266 on Build GRCh38
rs767326549 Chr.6: 35797324-35797325 on Build GRCh38
rs756767591 Chr.6: 35797299-35797299 on Build GRCh38
rs140966197 Chr.6: 35797224-35797224 on Build GRCh38
rs772312585 Chr.6: 35797271-35797271 on Build GRCh38
rs567706467 Chr.6: 35797340-35797340 on Build GRCh38
rs182775439 Chr.6: 35797310-35797310 on Build GRCh38
rs763805542 Chr.6: 35797241-35797242 on Build GRCh38
rs560934358 Chr.6: 35797241-35797241 on Build GRCh38
rs765589133 Chr.6: 35797292-35797292 on Build GRCh38
rs879535755 Chr.6: 35797290-35797291 on Build GRCh38
rs200437409 Chr.6: 35797247-35797247 on Build GRCh38
rs750392162 Chr.6: 35797301-35797301 on Build GRCh38
rs777015087 Chr.6: 35797220-35797220 on Build GRCh38
rs779758516 Chr.6: 35797314-35797314 on Build GRCh38
rs753617155 Chr.6: 35797315-35797315 on Build GRCh38
rs370676470 Chr.6: 35797196-35797196 on Build GRCh38
rs781207545 Chr.6: 35797255-35797255 on Build GRCh38
rs774377619 Chr.6: 35797184-35797184 on Build GRCh38
rs757789730 Chr.6: 35797170-35797170 on Build GRCh38
rs529464575 Chr.6: 35797258-35797258 on Build GRCh38
rs781246038 Chr.6: 35797336-35797336 on Build GRCh38
rs751060585 Chr.6: 35797265-35797273 on Build GRCh38
rs767121014 Chr.6: 35797300-35797300 on Build GRCh38
rs145002055 Chr.6: 35797229-35797229 on Build GRCh38
rs897962921 Chr.6: 35797307-35797307 on Build GRCh38
rs779892659 Chr.6: 35797263-35797263 on Build GRCh38
rs763798105 Chr.6: 35797297-35797297 on Build GRCh38
rs556188601 Chr.6: 35797139-35797139 on Build GRCh38
rs991604671 Chr.6: 35797150-35797150 on Build GRCh38
rs867356635 Chr.6: 35797228-35797228 on Build GRCh38
rs762528702 Chr.6: 35797224-35797225 on Build GRCh38
rs773306828 Chr.6: 35797274-35797274 on Build GRCh38

View Full Product Details