Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133355356-133355627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUOM
Entrez Gene ID
282969
Gene Name
fucose mutarotase
Gene Aliases
C10orf125, FUCU, FucM
UniGene
Hs.155823
Gene Chromosome Location
Chr.10: 133352773-133358031 on Build GRCh38
Transcripts
NM_001098483.2 NM_001301827.1 NM_001301828.1 NM_198472.2 XM_011539670.2 XM_011539671.2 XM_011539672.2 XM_011539673.2 XM_011539674.2 XM_011539675.2 XM_017016094.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4774618 c.446C>T p.A149V Chr.10: 133355389-133355389 on Build GRCh38
6419773 c.400G>T p.E134* Chr.10: 133355435-133355435 on Build GRCh38
5584586 c.455C>T p.P152L Chr.10: 133355380-133355380 on Build GRCh38
916500 c.460C>T p.L154L Chr.10: 133355375-133355375 on Build GRCh38
1194724 c.424A>C p.I142L Chr.10: 133355411-133355411 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000589020 Chr.10: 133355566-133355566 on Build GRCh38
rs779059728 Chr.10: 133355536-133355536 on Build GRCh38
rs757905897 Chr.10: 133355488-133355488 on Build GRCh38
rs761092586 Chr.10: 133355463-133355463 on Build GRCh38
rs550314591 Chr.10: 133355574-133355574 on Build GRCh38
rs762864791 Chr.10: 133355487-133355487 on Build GRCh38
rs115834264 Chr.10: 133355598-133355598 on Build GRCh38
rs756657550 Chr.10: 133355540-133355540 on Build GRCh38
rs372410658 Chr.10: 133355430-133355430 on Build GRCh38
rs931389187 Chr.10: 133355425-133355425 on Build GRCh38
rs766904028 Chr.10: 133355469-133355469 on Build GRCh38
rs371309419 Chr.10: 133355478-133355478 on Build GRCh38
rs867655798 Chr.10: 133355435-133355435 on Build GRCh38
rs756997396 Chr.10: 133355506-133355506 on Build GRCh38
rs1052285184 Chr.10: 133355569-133355570 on Build GRCh38
rs184108406 Chr.10: 133355431-133355431 on Build GRCh38
rs772321257 Chr.10: 133355548-133355548 on Build GRCh38
rs376896062 Chr.10: 133355460-133355460 on Build GRCh38
rs1043149985 Chr.10: 133355575-133355575 on Build GRCh38
rs371013369 Chr.10: 133355436-133355436 on Build GRCh38
rs758362565 Chr.10: 133355398-133355398 on Build GRCh38
rs754428093 Chr.10: 133355381-133355381 on Build GRCh38
rs201940330 Chr.10: 133355399-133355399 on Build GRCh38
rs913750931 Chr.10: 133355570-133355570 on Build GRCh38
rs773925502 Chr.10: 133355445-133355445 on Build GRCh38
rs753589954 Chr.10: 133355378-133355378 on Build GRCh38
rs370314829 Chr.10: 133355377-133355377 on Build GRCh38
rs772364371 Chr.10: 133355525-133355525 on Build GRCh38
rs374104109 Chr.10: 133355538-133355538 on Build GRCh38
rs953075085 Chr.10: 133355481-133355481 on Build GRCh38
rs752538305 Chr.10: 133355393-133355393 on Build GRCh38
rs748543014 Chr.10: 133355438-133355438 on Build GRCh38
rs368285198 Chr.10: 133355514-133355514 on Build GRCh38
rs965235571 Chr.10: 133355600-133355600 on Build GRCh38
rs960314142 Chr.10: 133355415-133355415 on Build GRCh38
rs77880897 Chr.10: 133355482-133355482 on Build GRCh38
rs1006495571 Chr.10: 133355585-133355585 on Build GRCh38
rs771102867 Chr.10: 133355467-133355467 on Build GRCh38
rs117818301 Chr.10: 133355576-133355576 on Build GRCh38
rs748174044 Chr.10: 133355457-133355457 on Build GRCh38
rs780983033 Chr.10: 133355441-133355441 on Build GRCh38
rs772689885 Chr.10: 133355439-133355439 on Build GRCh38
rs533342463 Chr.10: 133355483-133355483 on Build GRCh38
rs201184492 Chr.10: 133355411-133355411 on Build GRCh38
rs751545188 Chr.10: 133355492-133355492 on Build GRCh38
rs776378024 Chr.10: 133355476-133355476 on Build GRCh38
rs370634503 Chr.10: 133355523-133355523 on Build GRCh38
rs373850137 Chr.10: 133355420-133355420 on Build GRCh38
rs764825709 Chr.10: 133355391-133355391 on Build GRCh38

View Full Product Details