Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200857513-200857770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK1
Entrez Gene ID
1195
Gene Name
CDC like kinase 1
Gene Aliases
CLK, CLK/STY, STY
UniGene
Hs.433732
Gene Chromosome Location
Chr.2: 200853009-200864744 on Build GRCh38
Transcripts
NM_001162407.1 NM_004071.3 NR_027855.1 NR_027856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1482618 c.937C>G p.Q313E Chr.2: 200857739-200857739 on Build GRCh38
4945684 c.792T>G p.H264Q Chr.2: 200857758-200857758 on Build GRCh38
6156615 c.944G>T p.C315F Chr.2: 200857732-200857732 on Build GRCh38
5395912 c.910C>T p.L304L Chr.2: 200857766-200857766 on Build GRCh38
442099 c.811C>G p.Q271E Chr.2: 200857739-200857739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372875143 Chr.2: 200857697-200857697 on Build GRCh38
rs759733911 Chr.2: 200857689-200857689 on Build GRCh38
rs147232813 Chr.2: 200857725-200857725 on Build GRCh38
rs774453008 Chr.2: 200857596-200857596 on Build GRCh38
rs148758882 Chr.2: 200857626-200857626 on Build GRCh38
rs757838932 Chr.2: 200857704-200857704 on Build GRCh38
rs371057577 Chr.2: 200857737-200857737 on Build GRCh38
rs1025080991 Chr.2: 200857558-200857558 on Build GRCh38
rs752557590 Chr.2: 200857722-200857723 on Build GRCh38
rs771993715 Chr.2: 200857668-200857668 on Build GRCh38
rs756073686 Chr.2: 200857737-200857738 on Build GRCh38
rs12329121 Chr.2: 200857597-200857597 on Build GRCh38
rs760456006 Chr.2: 200857670-200857670 on Build GRCh38
rs932332808 Chr.2: 200857620-200857620 on Build GRCh38
rs990104833 Chr.2: 200857604-200857604 on Build GRCh38
rs909271062 Chr.2: 200857666-200857666 on Build GRCh38
rs376338724 Chr.2: 200857696-200857696 on Build GRCh38
rs370569142 Chr.2: 200857710-200857710 on Build GRCh38
rs924405577 Chr.2: 200857582-200857582 on Build GRCh38
rs570198667 Chr.2: 200857638-200857638 on Build GRCh38
rs533415389 Chr.2: 200857612-200857612 on Build GRCh38
rs370430530 Chr.2: 200857707-200857707 on Build GRCh38
rs747632604 Chr.2: 200857720-200857720 on Build GRCh38
rs144338593 Chr.2: 200857568-200857568 on Build GRCh38
rs972667325 Chr.2: 200857602-200857602 on Build GRCh38
rs776471227 Chr.2: 200857682-200857682 on Build GRCh38
rs773040465 Chr.2: 200857669-200857669 on Build GRCh38
rs746909965 Chr.2: 200857729-200857729 on Build GRCh38
rs967173874 Chr.2: 200857570-200857570 on Build GRCh38
rs770907467 Chr.2: 200857738-200857738 on Build GRCh38
rs746037806 Chr.2: 200857656-200857656 on Build GRCh38
rs188108176 Chr.2: 200857590-200857590 on Build GRCh38
rs902214342 Chr.2: 200857703-200857703 on Build GRCh38
rs912480915 Chr.2: 200857594-200857594 on Build GRCh38
rs373674851 Chr.2: 200857728-200857728 on Build GRCh38
rs770711146 Chr.2: 200857677-200857677 on Build GRCh38
rs977294438 Chr.2: 200857577-200857577 on Build GRCh38
rs2301982 Chr.2: 200857708-200857708 on Build GRCh38

View Full Product Details