Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6239485-6239746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA4
Entrez Gene ID
1262
Gene Name
cyclic nucleotide gated channel alpha 4
Gene Aliases
CNCA2, CNG-4, CNG4, CNG5, CNGB2, OCNC2, OCNCBETA, OCNCb
UniGene
Hs.434618
Gene Chromosome Location
Chr.11: 6234774-6245924 on Build GRCh38
Transcripts
NM_001037329.3 XM_017017217.1 XM_017017218.1 XM_017017219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5556679 c.189C>T p.H63H Chr.11: 6239708-6239708 on Build GRCh38
4529881 c.164G>A p.R55K Chr.11: 6239485-6239485 on Build GRCh38
6069822 c.225G>T p.T75T Chr.11: 6239744-6239744 on Build GRCh38
2039864 c.164+10C>T p.? Chr.11: 6239495-6239495 on Build GRCh38
3809889 c.190G>A p.G64S Chr.11: 6239709-6239709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951734834 Chr.11: 6239559-6239559 on Build GRCh38
rs993168983 Chr.11: 6239549-6239549 on Build GRCh38
rs753395347 Chr.11: 6239708-6239708 on Build GRCh38
rs911038935 Chr.11: 6239662-6239662 on Build GRCh38
rs759955658 Chr.11: 6239709-6239709 on Build GRCh38
rs547829254 Chr.11: 6239630-6239630 on Build GRCh38
rs773492976 Chr.11: 6239696-6239696 on Build GRCh38
rs540150885 Chr.11: 6239571-6239572 on Build GRCh38
rs972017863 Chr.11: 6239610-6239610 on Build GRCh38
rs781760773 Chr.11: 6239637-6239637 on Build GRCh38
rs971202902 Chr.11: 6239615-6239615 on Build GRCh38
rs1012687486 Chr.11: 6239562-6239562 on Build GRCh38
rs769812039 Chr.11: 6239684-6239684 on Build GRCh38
rs139738511 Chr.11: 6239555-6239555 on Build GRCh38
rs761908936 Chr.11: 6239681-6239681 on Build GRCh38
rs770371702 Chr.11: 6239507-6239507 on Build GRCh38
rs776515867 Chr.11: 6239679-6239679 on Build GRCh38
rs745462263 Chr.11: 6239648-6239648 on Build GRCh38
rs753216603 Chr.11: 6239720-6239720 on Build GRCh38
rs750569738 Chr.11: 6239511-6239511 on Build GRCh38
rs143078699 Chr.11: 6239609-6239609 on Build GRCh38
rs923281179 Chr.11: 6239629-6239629 on Build GRCh38
rs914594085 Chr.11: 6239654-6239654 on Build GRCh38
rs771329884 Chr.11: 6239620-6239620 on Build GRCh38
rs373506746 Chr.11: 6239520-6239520 on Build GRCh38
rs756798204 Chr.11: 6239642-6239642 on Build GRCh38
rs766591273 Chr.11: 6239634-6239634 on Build GRCh38
rs746805196 Chr.11: 6239663-6239663 on Build GRCh38
rs768640369 Chr.11: 6239676-6239676 on Build GRCh38
rs201459029 Chr.11: 6239635-6239635 on Build GRCh38
rs771918312 Chr.11: 6239651-6239651 on Build GRCh38
rs779650308 Chr.11: 6239652-6239652 on Build GRCh38
rs370908460 Chr.11: 6239638-6239638 on Build GRCh38
rs778659850 Chr.11: 6239645-6239645 on Build GRCh38
rs768180337 Chr.11: 6239715-6239715 on Build GRCh38
rs897439656 Chr.11: 6239518-6239518 on Build GRCh38
rs370277883 Chr.11: 6239697-6239697 on Build GRCh38

View Full Product Details