Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140561644-140561877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB3
Entrez Gene ID
10307
Gene Name
amyloid beta precursor protein binding family B member 3
Gene Aliases
FE65L2, SRA
UniGene
Hs.529449
Gene Chromosome Location
Chr.5: 140558268-140564604 on Build GRCh38
Transcripts
NM_006051.3 NM_133172.2 NM_133173.2 NM_133174.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754015 c.682G>C p.D228H Chr.5: 140561673-140561673 on Build GRCh38
3850669 c.687C>T p.S229S Chr.5: 140561668-140561668 on Build GRCh38
3946901 c.627-9C>G p.? Chr.5: 140561858-140561858 on Build GRCh38
6836576 c.701G>A p.C234Y Chr.5: 140561654-140561654 on Build GRCh38
7208379 c.683A>T p.D228V Chr.5: 140561672-140561672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770870465 Chr.5: 140561843-140561843 on Build GRCh38
rs1026550786 Chr.5: 140561796-140561796 on Build GRCh38
rs994997945 Chr.5: 140561755-140561755 on Build GRCh38
rs772242715 Chr.5: 140561743-140561743 on Build GRCh38
rs770958542 Chr.5: 140561728-140561728 on Build GRCh38
rs202146822 Chr.5: 140561832-140561832 on Build GRCh38
rs751041346 Chr.5: 140561797-140561797 on Build GRCh38
rs368784554 Chr.5: 140561814-140561814 on Build GRCh38
rs746000179 Chr.5: 140561856-140561856 on Build GRCh38
rs369537645 Chr.5: 140561725-140561725 on Build GRCh38
rs920094965 Chr.5: 140561675-140561675 on Build GRCh38
rs777740138 Chr.5: 140561711-140561711 on Build GRCh38
rs754347083 Chr.5: 140561756-140561756 on Build GRCh38
rs375471745 Chr.5: 140561758-140561758 on Build GRCh38
rs185656846 Chr.5: 140561710-140561710 on Build GRCh38
rs370991124 Chr.5: 140561846-140561846 on Build GRCh38
rs73267684 Chr.5: 140561828-140561828 on Build GRCh38
rs171632 Chr.5: 140561733-140561733 on Build GRCh38
rs200257140 Chr.5: 140561816-140561816 on Build GRCh38
rs950937591 Chr.5: 140561799-140561799 on Build GRCh38
rs779728971 Chr.5: 140561671-140561671 on Build GRCh38
rs759746853 Chr.5: 140561739-140561739 on Build GRCh38
rs749250621 Chr.5: 140561722-140561722 on Build GRCh38
rs752518398 Chr.5: 140561825-140561825 on Build GRCh38
rs765916942 Chr.5: 140561776-140561776 on Build GRCh38
rs372498339 Chr.5: 140561694-140561694 on Build GRCh38
rs747962593 Chr.5: 140561695-140561695 on Build GRCh38
rs201687337 Chr.5: 140561857-140561857 on Build GRCh38
rs368483411 Chr.5: 140561779-140561779 on Build GRCh38
rs764675208 Chr.5: 140561754-140561754 on Build GRCh38
rs760983951 Chr.5: 140561752-140561752 on Build GRCh38
rs867530450 Chr.5: 140561701-140561701 on Build GRCh38
rs752953242 Chr.5: 140561839-140561839 on Build GRCh38
rs377510549 Chr.5: 140561831-140561831 on Build GRCh38
rs147511887 Chr.5: 140561688-140561688 on Build GRCh38
rs754643630 Chr.5: 140561801-140561801 on Build GRCh38
rs754587835 Chr.5: 140561693-140561693 on Build GRCh38
rs775917575 Chr.5: 140561748-140561748 on Build GRCh38
rs34020914 Chr.5: 140561728-140561729 on Build GRCh38

View Full Product Details