Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55177713-55177938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLHC1
Entrez Gene ID
130162
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
C2orf63
UniGene
Hs.347014
Gene Chromosome Location
Chr.2: 55172547-55232563 on Build GRCh38
Transcripts
NM_001135598.1 NM_152385.2 XM_005264136.2 XM_006711937.3 XM_006711938.3 XM_006711939.3 XM_011532519.2 XM_011532520.2 XM_011532521.2 XM_011532522.2 XM_011532523.2 XM_011532524.1 XM_011532525.2 XM_011532527.2 XM_011532528.1 XM_011532529.2 XM_011532530.1 XM_011532534.1 XM_011532535.2 XM_017003333.1 XM_017003334.1 XM_017003335.1 XM_017003336.1 XM_017003337.1 XM_017003338.1 XM_017003339.1 XM_017003340.1 XM_017003341.1 XM_017003342.1 XM_017003343.1 XM_017003344.1 XM_017003345.1 XM_017003346.1 XM_017003347.1 XM_017003349.1 XM_017003350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397603 c.1412G>T p.C471F Chr.2: 55177754-55177754 on Build GRCh38
6394688 c.1407G>T p.M469I Chr.2: 55177759-55177759 on Build GRCh38
6542650 c.1423G>A p.E475K Chr.2: 55177743-55177743 on Build GRCh38
6387835 c.1398G>T p.Q466H Chr.2: 55177768-55177768 on Build GRCh38
148860 c.1419A>G p.Q473Q Chr.2: 55177747-55177747 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751030292 Chr.2: 55177796-55177796 on Build GRCh38
rs750347831 Chr.2: 55177805-55177805 on Build GRCh38
rs6545477 Chr.2: 55177789-55177789 on Build GRCh38
rs760712433 Chr.2: 55177827-55177827 on Build GRCh38
rs997999341 Chr.2: 55177808-55177808 on Build GRCh38
rs763541968 Chr.2: 55177794-55177794 on Build GRCh38
rs762632306 Chr.2: 55177790-55177790 on Build GRCh38
rs1049626600 Chr.2: 55177840-55177840 on Build GRCh38
rs75442422 Chr.2: 55177901-55177901 on Build GRCh38
rs372061125 Chr.2: 55177783-55177783 on Build GRCh38
rs13032294 Chr.2: 55177747-55177747 on Build GRCh38
rs772653130 Chr.2: 55177767-55177767 on Build GRCh38
rs768659419 Chr.2: 55177757-55177757 on Build GRCh38
rs572892152 Chr.2: 55177756-55177756 on Build GRCh38
rs200921597 Chr.2: 55177803-55177803 on Build GRCh38
rs945333940 Chr.2: 55177792-55177792 on Build GRCh38
rs773720803 Chr.2: 55177773-55177773 on Build GRCh38
rs937641242 Chr.2: 55177866-55177866 on Build GRCh38
rs889700577 Chr.2: 55177869-55177869 on Build GRCh38
rs749504729 Chr.2: 55177811-55177811 on Build GRCh38
rs533665450 Chr.2: 55177813-55177813 on Build GRCh38
rs755951708 Chr.2: 55177806-55177806 on Build GRCh38
rs896229122 Chr.2: 55177909-55177909 on Build GRCh38
rs746077380 Chr.2: 55177881-55177881 on Build GRCh38
rs779795629 Chr.2: 55177807-55177807 on Build GRCh38
rs781289849 Chr.2: 55177797-55177798 on Build GRCh38
rs774452457 Chr.2: 55177762-55177762 on Build GRCh38
rs748207070 Chr.2: 55177826-55177826 on Build GRCh38
rs181837553 Chr.2: 55177908-55177908 on Build GRCh38
rs766832824 Chr.2: 55177778-55177778 on Build GRCh38
rs778874153 Chr.2: 55177815-55177815 on Build GRCh38
rs748627006 Chr.2: 55177765-55177765 on Build GRCh38
rs748196506 Chr.2: 55177819-55177819 on Build GRCh38
rs761184708 Chr.2: 55177774-55177774 on Build GRCh38
rs542937675 Chr.2: 55177912-55177912 on Build GRCh38

View Full Product Details