Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103086545-103086786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC10
Entrez Gene ID
83787
Gene Name
armadillo repeat containing 10
Gene Aliases
PNAS-112, PNAS112, PSEC0198, SVH
UniGene
Hs.287412
Gene Chromosome Location
Chr.7: 103074727-103099763 on Build GRCh38
Transcripts
NM_001161009.2 NM_001161010.2 NM_001161011.2 NM_001161012.2 NM_001161013.2 NM_031905.4 XM_005250628.2 XM_011516601.2 XM_011516602.2 XM_017012684.1 XM_017012685.1 XM_017012686.1 XM_017012687.1 XM_017012688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1083534 c.468G>T p.E156D Chr.7: 103086704-103086704 on Build GRCh38
305469 c.495G>C p.L165L Chr.7: 103086731-103086731 on Build GRCh38
4449963 c.394-1G>A p.? Chr.7: 103086629-103086629 on Build GRCh38
6378163 c.445C>A p.H149N Chr.7: 103086681-103086681 on Build GRCh38
4759578 c.521A>C p.K174T Chr.7: 103086757-103086757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138429012 Chr.7: 103086640-103086640 on Build GRCh38
rs961778255 Chr.7: 103086734-103086734 on Build GRCh38
rs767354280 Chr.7: 103086582-103086582 on Build GRCh38
rs369209750 Chr.7: 103086605-103086605 on Build GRCh38
rs764185097 Chr.7: 103086698-103086699 on Build GRCh38
rs780692148 Chr.7: 103086625-103086625 on Build GRCh38
rs752848262 Chr.7: 103086619-103086620 on Build GRCh38
rs775335105 Chr.7: 103086675-103086675 on Build GRCh38
rs77388570 Chr.7: 103086623-103086623 on Build GRCh38
rs748266889 Chr.7: 103086642-103086642 on Build GRCh38
rs1020692606 Chr.7: 103086746-103086746 on Build GRCh38
rs906438789 Chr.7: 103086615-103086615 on Build GRCh38
rs763890216 Chr.7: 103086588-103086588 on Build GRCh38
rs754001002 Chr.7: 103086620-103086620 on Build GRCh38
rs771314782 Chr.7: 103086605-103086606 on Build GRCh38
rs751966274 Chr.7: 103086692-103086692 on Build GRCh38
rs534736196 Chr.7: 103086605-103086607 on Build GRCh38
rs755814530 Chr.7: 103086586-103086586 on Build GRCh38
rs887500872 Chr.7: 103086600-103086600 on Build GRCh38
rs753602156 Chr.7: 103086684-103086684 on Build GRCh38
rs202066616 Chr.7: 103086661-103086661 on Build GRCh38
rs199625691 Chr.7: 103086585-103086585 on Build GRCh38
rs747564739 Chr.7: 103086627-103086627 on Build GRCh38
rs751569484 Chr.7: 103086719-103086721 on Build GRCh38
rs373032426 Chr.7: 103086626-103086626 on Build GRCh38
rs755527087 Chr.7: 103086716-103086716 on Build GRCh38
rs763113965 Chr.7: 103086690-103086691 on Build GRCh38
rs759878887 Chr.7: 103086604-103086605 on Build GRCh38
rs935931064 Chr.7: 103086575-103086575 on Build GRCh38
rs199803039 Chr.7: 103086650-103086650 on Build GRCh38
rs761348176 Chr.7: 103086685-103086685 on Build GRCh38
rs777916108 Chr.7: 103086733-103086733 on Build GRCh38
rs535555977 Chr.7: 103086647-103086647 on Build GRCh38
rs753072526 Chr.7: 103086720-103086720 on Build GRCh38
rs759549028 Chr.7: 103086660-103086660 on Build GRCh38
rs756892375 Chr.7: 103086599-103086599 on Build GRCh38
rs75142391 Chr.7: 103086619-103086619 on Build GRCh38
rs76904063 Chr.7: 103086704-103086704 on Build GRCh38
rs867678070 Chr.7: 103086728-103086728 on Build GRCh38
rs755230564 Chr.7: 103086633-103086633 on Build GRCh38
rs752252428 Chr.7: 103086584-103086584 on Build GRCh38
rs534592982 Chr.7: 103086576-103086576 on Build GRCh38
rs1014592815 Chr.7: 103086724-103086724 on Build GRCh38
rs760428556 Chr.7: 103086677-103086677 on Build GRCh38
rs778039256 Chr.7: 103086649-103086649 on Build GRCh38
rs150220068 Chr.7: 103086731-103086731 on Build GRCh38
rs764844689 Chr.7: 103086690-103086690 on Build GRCh38
rs753369485 Chr.7: 103086594-103086594 on Build GRCh38
rs781595166 Chr.7: 103086719-103086719 on Build GRCh38
rs770958525 Chr.7: 103086750-103086750 on Build GRCh38
rs369144824 Chr.7: 103086725-103086725 on Build GRCh38
rs749517792 Chr.7: 103086735-103086735 on Build GRCh38
rs771015681 Chr.7: 103086657-103086657 on Build GRCh38
rs902285154 Chr.7: 103086751-103086751 on Build GRCh38

View Full Product Details