Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7730058-7730287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4G
Entrez Gene ID
339390
Gene Name
C-type lectin domain family 4 member G
Gene Aliases
DTTR431, LP2698, LSECtin, UNQ431
UniGene
Hs.220649
Gene Chromosome Location
Chr.19: 7728957-7732171 on Build GRCh38
Transcripts
NM_001244856.1 NM_198492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5576536 c.566C>T p.A189V Chr.19: 7730080-7730080 on Build GRCh38
4749521 c.517G>A p.E173K Chr.19: 7730129-7730129 on Build GRCh38
6630203 c.497C>T p.P166L Chr.19: 7730149-7730149 on Build GRCh38
6630204 c.579C>T p.C193C Chr.19: 7730067-7730067 on Build GRCh38
4450183 c.479-1G>T p.? Chr.19: 7730168-7730168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778258404 Chr.19: 7730132-7730132 on Build GRCh38
rs749863098 Chr.19: 7730191-7730191 on Build GRCh38
rs368297239 Chr.19: 7730091-7730091 on Build GRCh38
rs149297990 Chr.19: 7730172-7730172 on Build GRCh38
rs781657449 Chr.19: 7730109-7730109 on Build GRCh38
rs756733681 Chr.19: 7730195-7730195 on Build GRCh38
rs755178489 Chr.19: 7730164-7730164 on Build GRCh38
rs760711933 Chr.19: 7730079-7730079 on Build GRCh38
rs756487039 Chr.19: 7730173-7730173 on Build GRCh38
rs760415857 Chr.19: 7730154-7730154 on Build GRCh38
rs763109759 Chr.19: 7730159-7730159 on Build GRCh38
rs778243626 Chr.19: 7730183-7730183 on Build GRCh38
rs1014143982 Chr.19: 7730175-7730175 on Build GRCh38
rs891996050 Chr.19: 7730176-7730176 on Build GRCh38
rs565957284 Chr.19: 7730137-7730137 on Build GRCh38
rs980811058 Chr.19: 7730242-7730242 on Build GRCh38
rs773992274 Chr.19: 7730143-7730143 on Build GRCh38
rs201996252 Chr.19: 7730083-7730083 on Build GRCh38
rs555610194 Chr.19: 7730106-7730106 on Build GRCh38
rs776721083 Chr.19: 7730157-7730157 on Build GRCh38
rs745578277 Chr.19: 7730148-7730148 on Build GRCh38
rs200560949 Chr.19: 7730209-7730209 on Build GRCh38
rs766631531 Chr.19: 7730160-7730160 on Build GRCh38
rs758116896 Chr.19: 7730246-7730246 on Build GRCh38
rs756574920 Chr.19: 7730126-7730126 on Build GRCh38
rs566389972 Chr.19: 7730145-7730145 on Build GRCh38
rs958030782 Chr.19: 7730269-7730270 on Build GRCh38
rs748483954 Chr.19: 7730125-7730125 on Build GRCh38
rs770488586 Chr.19: 7730139-7730139 on Build GRCh38
rs949740120 Chr.19: 7730263-7730263 on Build GRCh38
rs775302612 Chr.19: 7730151-7730151 on Build GRCh38
rs750487345 Chr.19: 7730086-7730086 on Build GRCh38
rs771737816 Chr.19: 7730149-7730149 on Build GRCh38
rs1010346889 Chr.19: 7730185-7730185 on Build GRCh38
rs558554651 Chr.19: 7730251-7730251 on Build GRCh38
rs772952991 Chr.19: 7730187-7730188 on Build GRCh38
rs972725268 Chr.19: 7730248-7730248 on Build GRCh38
rs751865095 Chr.19: 7730104-7730104 on Build GRCh38
rs969470009 Chr.19: 7730216-7730216 on Build GRCh38
rs767665694 Chr.19: 7730166-7730166 on Build GRCh38

View Full Product Details