Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127892611-127892866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLRN3
Entrez Gene ID
119467
Gene Name
clarin 3
Gene Aliases
TMEM12, USH3AL1
UniGene
Hs.242014
Gene Chromosome Location
Chr.10: 127877841-127892947 on Build GRCh38
Transcripts
NM_152311.3 XM_011539274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806727 c.154A>G p.T52A Chr.10: 127892631-127892631 on Build GRCh38
5781852 c.7A>G p.T3A Chr.10: 127892778-127892778 on Build GRCh38
4573469 c.107G>A p.S36N Chr.10: 127892678-127892678 on Build GRCh38
5948953 c.114T>G p.I38M Chr.10: 127892671-127892671 on Build GRCh38
4471093 c.169C>T p.R57C Chr.10: 127892616-127892616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535687958 Chr.10: 127892678-127892678 on Build GRCh38
rs751070162 Chr.10: 127892659-127892659 on Build GRCh38
rs867290114 Chr.10: 127892699-127892699 on Build GRCh38
rs866390748 Chr.10: 127892808-127892808 on Build GRCh38
rs565998993 Chr.10: 127892725-127892725 on Build GRCh38
rs921024138 Chr.10: 127892777-127892777 on Build GRCh38
rs778756901 Chr.10: 127892823-127892823 on Build GRCh38
rs200762118 Chr.10: 127892683-127892683 on Build GRCh38
rs1048493536 Chr.10: 127892634-127892634 on Build GRCh38
rs756845457 Chr.10: 127892663-127892663 on Build GRCh38
rs553761084 Chr.10: 127892703-127892703 on Build GRCh38
rs776221225 Chr.10: 127892812-127892813 on Build GRCh38
rs764642712 Chr.10: 127892743-127892743 on Build GRCh38
rs754781454 Chr.10: 127892813-127892813 on Build GRCh38
rs372098394 Chr.10: 127892805-127892805 on Build GRCh38
rs765500931 Chr.10: 127892733-127892733 on Build GRCh38
rs1015784565 Chr.10: 127892736-127892736 on Build GRCh38
rs771049433 Chr.10: 127892720-127892720 on Build GRCh38
rs753157078 Chr.10: 127892734-127892734 on Build GRCh38
rs17852659 Chr.10: 127892674-127892674 on Build GRCh38
rs750028999 Chr.10: 127892792-127892792 on Build GRCh38
rs770771690 Chr.10: 127892813-127892813 on Build GRCh38
rs769470783 Chr.10: 127892688-127892688 on Build GRCh38
rs111836033 Chr.10: 127892633-127892633 on Build GRCh38
rs932356239 Chr.10: 127892781-127892781 on Build GRCh38
rs3802693 Chr.10: 127892807-127892807 on Build GRCh38
rs774294383 Chr.10: 127892715-127892715 on Build GRCh38
rs776716340 Chr.10: 127892723-127892723 on Build GRCh38
rs780647822 Chr.10: 127892667-127892667 on Build GRCh38
rs367866010 Chr.10: 127892770-127892770 on Build GRCh38
rs956068740 Chr.10: 127892776-127892776 on Build GRCh38
rs137990791 Chr.10: 127892827-127892827 on Build GRCh38
rs763446777 Chr.10: 127892735-127892735 on Build GRCh38
rs557709645 Chr.10: 127892782-127892782 on Build GRCh38
rs755838938 Chr.10: 127892801-127892801 on Build GRCh38
rs749033260 Chr.10: 127892811-127892811 on Build GRCh38
rs983017429 Chr.10: 127892649-127892649 on Build GRCh38
rs539370192 Chr.10: 127892751-127892751 on Build GRCh38
rs375846400 Chr.10: 127892696-127892696 on Build GRCh38
rs746838069 Chr.10: 127892803-127892804 on Build GRCh38
rs908228654 Chr.10: 127892635-127892635 on Build GRCh38
rs748152154 Chr.10: 127892832-127892832 on Build GRCh38
rs939892579 Chr.10: 127892639-127892639 on Build GRCh38
rs116562518 Chr.10: 127892648-127892648 on Build GRCh38
rs768619796 Chr.10: 127892713-127892713 on Build GRCh38
rs770904095 Chr.10: 127892834-127892834 on Build GRCh38
rs895634972 Chr.10: 127892651-127892651 on Build GRCh38
rs1044479594 Chr.10: 127892687-127892687 on Build GRCh38
rs778110517 Chr.10: 127892740-127892741 on Build GRCh38
rs760690107 Chr.10: 127892716-127892716 on Build GRCh38

View Full Product Details