Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 82639277-82639550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH13
Entrez Gene ID
1012
Gene Name
cadherin 13
Gene Aliases
CDHH, P105
UniGene
Hs.654386
Gene Chromosome Location
Chr.16: 82626794-83796610 on Build GRCh38
Transcripts
NM_001220488.1 NM_001220489.1 NM_001220490.1 NM_001220491.1 NM_001220492.1 NM_001257.4 XM_017022848.1 XM_017022849.1

SNP Detail

SNP ID Chromosome Location
rs370056914 Chr.16: 82639298-82639298 on Build GRCh38
rs950705310 Chr.16: 82639375-82639375 on Build GRCh38
rs1019561501 Chr.16: 82639344-82639344 on Build GRCh38
rs184153489 Chr.16: 82639494-82639494 on Build GRCh38
rs764679680 Chr.16: 82639313-82639313 on Build GRCh38
rs569316960 Chr.16: 82639515-82639515 on Build GRCh38
rs80112483 Chr.16: 82639345-82639345 on Build GRCh38
rs565426910 Chr.16: 82639488-82639488 on Build GRCh38
rs954023319 Chr.16: 82639336-82639336 on Build GRCh38
rs1026049645 Chr.16: 82639354-82639354 on Build GRCh38
rs1039836467 Chr.16: 82639476-82639476 on Build GRCh38
rs1026867834 Chr.16: 82639529-82639529 on Build GRCh38
rs750845699 Chr.16: 82639384-82639384 on Build GRCh38
rs558256631 Chr.16: 82639362-82639362 on Build GRCh38
rs945699483 Chr.16: 82639449-82639449 on Build GRCh38
rs1051786620 Chr.16: 82639462-82639462 on Build GRCh38
rs919904025 Chr.16: 82639357-82639357 on Build GRCh38
rs991320229 Chr.16: 82639407-82639407 on Build GRCh38
rs1019446892 Chr.16: 82639511-82639511 on Build GRCh38
rs142193701 Chr.16: 82639316-82639316 on Build GRCh38
rs576747633 Chr.16: 82639466-82639466 on Build GRCh38
rs532537858 Chr.16: 82639348-82639348 on Build GRCh38
rs4782724 Chr.16: 82639442-82639442 on Build GRCh38
rs576859115 Chr.16: 82639403-82639403 on Build GRCh38
rs12444222 Chr.16: 82639315-82639315 on Build GRCh38
rs376170874 Chr.16: 82639437-82639437 on Build GRCh38
rs772477362 Chr.16: 82639463-82639463 on Build GRCh38
rs892076384 Chr.16: 82639299-82639299 on Build GRCh38
rs891297288 Chr.16: 82639330-82639330 on Build GRCh38
rs566006904 Chr.16: 82639307-82639307 on Build GRCh38
rs935723602 Chr.16: 82639421-82639421 on Build GRCh38
rs138964780 Chr.16: 82639314-82639314 on Build GRCh38
rs983763477 Chr.16: 82639376-82639376 on Build GRCh38
rs1053184658 Chr.16: 82639312-82639312 on Build GRCh38
rs115056752 Chr.16: 82639489-82639489 on Build GRCh38
rs550740563 Chr.16: 82639436-82639436 on Build GRCh38
rs927042269 Chr.16: 82639420-82639420 on Build GRCh38
rs1018334221 Chr.16: 82639338-82639338 on Build GRCh38
rs780660253 Chr.16: 82639405-82639405 on Build GRCh38
rs897099301 Chr.16: 82639513-82639513 on Build GRCh38
rs530243818 Chr.16: 82639521-82639521 on Build GRCh38
rs1034166222 Chr.16: 82639518-82639518 on Build GRCh38
rs150875489 Chr.16: 82639464-82639464 on Build GRCh38
rs7194373 Chr.16: 82639485-82639485 on Build GRCh38
rs1008027223 Chr.16: 82639339-82639339 on Build GRCh38
rs924223324 Chr.16: 82639446-82639446 on Build GRCh38
rs942885936 Chr.16: 82639475-82639475 on Build GRCh38
rs979907299 Chr.16: 82639418-82639418 on Build GRCh38
rs972759776 Chr.16: 82639352-82639352 on Build GRCh38
rs995497087 Chr.16: 82639349-82639349 on Build GRCh38
rs542924912 Chr.16: 82639461-82639461 on Build GRCh38
rs935565714 Chr.16: 82639456-82639456 on Build GRCh38
rs754321991 Chr.16: 82639435-82639435 on Build GRCh38

View Full Product Details