Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63309625-63309898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL20A1
Entrez Gene ID
57642
Gene Name
collagen type XX alpha 1 chain
Gene Aliases
bA261N11.4
UniGene
Hs.271285
Gene Chromosome Location
Chr.20: 63293169-63334847 on Build GRCh38
Transcripts
NM_020882.2 XM_011528937.1 XM_011528938.1 XM_011528939.1 XM_011528940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
359727 c.1118C>A p.A373D Chr.20: 63309770-63309770 on Build GRCh38
2931663 c.1125C>G p.D375E Chr.20: 63309777-63309777 on Build GRCh38
5180716 c.1239C>T p.A413A Chr.20: 63309891-63309891 on Build GRCh38
6526076 c.1259C>A p.A420D Chr.20: 63309890-63309890 on Build GRCh38
6526077 c.1238C>A p.A413D Chr.20: 63309890-63309890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768754881 Chr.20: 63309781-63309781 on Build GRCh38
rs986635188 Chr.20: 63309778-63309778 on Build GRCh38
rs1011546109 Chr.20: 63309850-63309850 on Build GRCh38
rs912376488 Chr.20: 63309793-63309793 on Build GRCh38
rs769946191 Chr.20: 63309798-63309798 on Build GRCh38
rs1025064451 Chr.20: 63309732-63309732 on Build GRCh38
rs778864152 Chr.20: 63309761-63309761 on Build GRCh38
rs34485195 Chr.20: 63309803-63309804 on Build GRCh38
rs923039373 Chr.20: 63309686-63309687 on Build GRCh38
rs773573190 Chr.20: 63309800-63309800 on Build GRCh38
rs186757692 Chr.20: 63309865-63309865 on Build GRCh38
rs763265567 Chr.20: 63309806-63309806 on Build GRCh38
rs374436824 Chr.20: 63309709-63309709 on Build GRCh38
rs377073867 Chr.20: 63309851-63309851 on Build GRCh38
rs752283267 Chr.20: 63309752-63309754 on Build GRCh38
rs766845299 Chr.20: 63309738-63309738 on Build GRCh38
rs777540579 Chr.20: 63309818-63309820 on Build GRCh38
rs368982947 Chr.20: 63309857-63309857 on Build GRCh38
rs377006506 Chr.20: 63309762-63309762 on Build GRCh38
rs774321889 Chr.20: 63309809-63309809 on Build GRCh38
rs776788181 Chr.20: 63309792-63309792 on Build GRCh38
rs1044526845 Chr.20: 63309787-63309787 on Build GRCh38
rs755838084 Chr.20: 63309824-63309824 on Build GRCh38
rs752451966 Chr.20: 63309822-63309822 on Build GRCh38
rs1055325742 Chr.20: 63309715-63309715 on Build GRCh38
rs200924087 Chr.20: 63309820-63309820 on Build GRCh38
rs763464712 Chr.20: 63309728-63309728 on Build GRCh38
rs1037457773 Chr.20: 63309688-63309688 on Build GRCh38
rs765585805 Chr.20: 63309717-63309717 on Build GRCh38
rs983326841 Chr.20: 63309860-63309860 on Build GRCh38
rs766815439 Chr.20: 63309807-63309807 on Build GRCh38
rs914454972 Chr.20: 63309649-63309649 on Build GRCh38
rs763769391 Chr.20: 63309832-63309832 on Build GRCh38
rs774897563 Chr.20: 63309811-63309811 on Build GRCh38
rs964604875 Chr.20: 63309879-63309879 on Build GRCh38
rs943195152 Chr.20: 63309654-63309654 on Build GRCh38
rs370160805 Chr.20: 63309810-63309810 on Build GRCh38
rs201122273 Chr.20: 63309744-63309744 on Build GRCh38
rs372246087 Chr.20: 63309713-63309713 on Build GRCh38
rs575454514 Chr.20: 63309685-63309685 on Build GRCh38
rs989813591 Chr.20: 63309749-63309749 on Build GRCh38
rs757779465 Chr.20: 63309770-63309770 on Build GRCh38
rs777692696 Chr.20: 63309746-63309746 on Build GRCh38
rs372427337 Chr.20: 63309752-63309752 on Build GRCh38
rs201491872 Chr.20: 63309833-63309833 on Build GRCh38
rs376834639 Chr.20: 63309726-63309726 on Build GRCh38
rs748358276 Chr.20: 63309794-63309794 on Build GRCh38
rs745817287 Chr.20: 63309856-63309856 on Build GRCh38
rs752366140 Chr.20: 63309745-63309745 on Build GRCh38
rs762209765 Chr.20: 63309716-63309716 on Build GRCh38
rs759007324 Chr.20: 63309812-63309812 on Build GRCh38
rs995901504 Chr.20: 63309706-63309706 on Build GRCh38
rs940083947 Chr.20: 63309677-63309677 on Build GRCh38
rs757055637 Chr.20: 63309848-63309848 on Build GRCh38
rs781346441 Chr.20: 63309861-63309861 on Build GRCh38
rs546168021 Chr.20: 63309701-63309701 on Build GRCh38

View Full Product Details