Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178834120-178834379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC141
Entrez Gene ID
285025
Gene Name
coiled-coil domain containing 141
Gene Aliases
CAMDI
UniGene
Hs.324341
Gene Chromosome Location
Chr.2: 178825308-179050134 on Build GRCh38
Transcripts
NM_173648.3 XM_011510991.2 XM_011510992.2 XM_011510993.2 XM_017003867.1 XM_017003868.1 XM_017003869.1 XM_017003870.1 XM_017003871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6510338 c.4578G>T p.W1526C Chr.2: 178834188-178834188 on Build GRCh38
5895187 c.4445_4446insC p.A1483fs*5 Chr.2: 178834320-178834321 on Build GRCh38
5630549 c.4460G>A p.S1487N Chr.2: 178834306-178834306 on Build GRCh38
4945743 c.4441G>T p.A1481S Chr.2: 178834325-178834325 on Build GRCh38
5941618 c.4457C>T p.S1486F Chr.2: 178834309-178834309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924885304 Chr.2: 178834170-178834170 on Build GRCh38
rs946058929 Chr.2: 178834205-178834205 on Build GRCh38
rs998023741 Chr.2: 178834268-178834268 on Build GRCh38
rs947613526 Chr.2: 178834153-178834153 on Build GRCh38
rs16866554 Chr.2: 178834218-178834218 on Build GRCh38
rs994534626 Chr.2: 178834280-178834280 on Build GRCh38
rs539116633 Chr.2: 178834329-178834329 on Build GRCh38
rs892049188 Chr.2: 178834201-178834201 on Build GRCh38
rs199670476 Chr.2: 178834304-178834304 on Build GRCh38
rs757410634 Chr.2: 178834145-178834145 on Build GRCh38
rs570537506 Chr.2: 178834321-178834321 on Build GRCh38
rs746733810 Chr.2: 178834234-178834234 on Build GRCh38
rs569278815 Chr.2: 178834352-178834352 on Build GRCh38
rs149753504 Chr.2: 178834158-178834158 on Build GRCh38
rs762178070 Chr.2: 178834344-178834344 on Build GRCh38
rs1005216478 Chr.2: 178834298-178834298 on Build GRCh38
rs199532854 Chr.2: 178834267-178834267 on Build GRCh38
rs370167073 Chr.2: 178834246-178834246 on Build GRCh38
rs1044062588 Chr.2: 178834241-178834241 on Build GRCh38
rs549611076 Chr.2: 178834190-178834190 on Build GRCh38
rs375249342 Chr.2: 178834322-178834322 on Build GRCh38
rs1057493257 Chr.2: 178834213-178834213 on Build GRCh38
rs779888503 Chr.2: 178834144-178834144 on Build GRCh38
rs779194018 Chr.2: 178834155-178834155 on Build GRCh38
rs1017036149 Chr.2: 178834301-178834301 on Build GRCh38
rs951410868 Chr.2: 178834293-178834294 on Build GRCh38
rs780755107 Chr.2: 178834270-178834270 on Build GRCh38
rs551674463 Chr.2: 178834305-178834305 on Build GRCh38
rs549389165 Chr.2: 178834335-178834335 on Build GRCh38
rs760885419 Chr.2: 178834340-178834340 on Build GRCh38
rs796601776 Chr.2: 178834187-178834187 on Build GRCh38
rs774449715 Chr.2: 178834217-178834217 on Build GRCh38
rs913480360 Chr.2: 178834349-178834349 on Build GRCh38
rs769278075 Chr.2: 178834302-178834302 on Build GRCh38
rs185082965 Chr.2: 178834274-178834274 on Build GRCh38
rs764624099 Chr.2: 178834341-178834341 on Build GRCh38
rs201233545 Chr.2: 178834173-178834173 on Build GRCh38
rs754101714 Chr.2: 178834343-178834343 on Build GRCh38

View Full Product Details